Skip to Content
Merck
All Photos(6)

Key Documents

178756

Sigma-Aldrich

Diphenyl phosphoryl azide

97%

Synonym(s):

DPPA, Phosphoric acid diphenyl ester azide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2P(O)N3
CAS Number:
Molecular Weight:
275.20
Beilstein:
2058967
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.551 (lit.)

bp

157 °C/0.17 mmHg (lit.)

density

1.277 g/mL at 25 °C (lit.)

functional group

azide

storage temp.

2-8°C

SMILES string

[N-]=[N+]=NP(=O)(Oc1ccccc1)Oc2ccccc2

InChI

1S/C12H10N3O3P/c13-14-15-19(16,17-11-7-3-1-4-8-11)18-12-9-5-2-6-10-12/h1-10H

InChI key

SORGEQQSQGNZFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydroazidation Catalyst for Facile Preparation of Organoazides

Reagent for synthesis of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement
Aziridination of olefins catalyzed by colbalt-tetraphenylporphyrin.
Used as the activating agent in the preparation of macrocyclic lactams and of an aldose reductase inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 31, 6469-6469 (1990)
The Journal of Organic Chemistry, 71, 6665-6665 (2006)
E Benqué et al.
Journal of clinical periodontology, 24(8), 544-549 (1997-08-01)
This study, confined to non-smokers, evaluated guided tissue regeneration using a diphenylphosphorylazide-cross-linked bovine type I collagen membrane in deep 3-wall intrabony defects in 52 adult periodontitis (AP) and 16-rapidly progressive periodontitis (RPP) patients, previously treated for the acute phase of
Marcus Baumann et al.
Organic & biomolecular chemistry, 6(9), 1577-1586 (2008-04-19)
The use of a mesofluidic flow reactor is described for performing Curtius rearrangement reactions of carboxylic acids in the presence of diphenylphosphoryl azide and trapping of the intermediate isocyanates with various nucleophiles.
S Zahedi et al.
Journal of periodontology, 69(9), 975-981 (1998-10-17)
To retard collagen membrane enzymatic degradation and to increase its mechanical strength, the diphenylphosphorylazide (DPPA) technique has been demonstrated to achieve natural cross-links between peptide chains of collagen without leaving any foreign product in the cross-linked molecule. In the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service