Skip to Content
Merck
All Photos(1)

Key Documents

467952

Sigma-Aldrich

1-Bromo-2-cyclohexylethane

98%

Synonym(s):

2-Cyclohexylethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH2Br
CAS Number:
Molecular Weight:
191.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.49 (lit.)

bp

209 °C (lit.)

density

1.221 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrCCC1CCCCC1

InChI

1S/C8H15Br/c9-7-6-8-4-2-1-3-5-8/h8H,1-7H2

InChI key

JRQAAYVLPPGEHT-UHFFFAOYSA-N

General description

1-Bromo-2-cyclohexylethane is a halogenated hydrocarbon. Reaction of 1-bromo-2-cyclohexylethane with pillar[5]arene has been investigated.

Application

1-Bromo-2-cyclohexylethane may be used in the synthesis of 2-cyclohexylethyloxy-4-nitroaniline.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Cragg et al.
Chemical Society reviews, 41(2), 597-607 (2011-08-02)
Pillar[5]arenes are [1(5)]paracyclophane derivatives consisting of 1,4-disubstituted hydroquinones linked by methylene bridges in the 2,5-positions. The first report of these novel macrocycles was in 2008, when 1,4-dimethoxypillar[5]arene was prepared in 22% yield, and subsequent improvements in synthetic methods have allowed
Isomerization of alkyl branched alkynoic acids.
Abrams S R.
Canadian Journal of Chemistry, 64(3), 457-463 (1986)
Min Wang et al.
Bioorganic & medicinal chemistry letters, 17(2), 332-336 (2006-11-11)
Aromatase is a particularly good target in the treatment of estrogen receptor positive breast cancer. Novel carbon-11 labeled sulfonanilide analogues, N-[11C]methyl-N-(2-alkyloxy-4-nitrophenyl)-methanesulfonamides ([11C]3a-f, alkyl=propyl, isopropyl, 1-ethyl-propyl, cyclopentyl, cyclohexyl, and cyclohexylethyl), were designed and synthesized as potential PET agents for imaging of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service