184136
11-Bromo-1-undecanol
98%
Synonym(s):
Undecamethylene bromohydrin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Br(CH2)11OH
CAS Number:
Molecular Weight:
251.20
Beilstein:
1741180
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
bp
165-170 °C/1 mmHg (lit.)
mp
46-49 °C (lit.)
solubility
chloroform: soluble 50 mg/mL, clear to slightly hazy, colorless to dark yellow
functional group
bromo
hydroxyl
SMILES string
OCCCCCCCCCCCBr
InChI
1S/C11H23BrO/c12-10-8-6-4-2-1-3-5-7-9-11-13/h13H,1-11H2
InChI key
XFGANBYCJWQYBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
11-Bromo-1-undecanol was used in the preparation of 11-mercapto-1-undecanol and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
One-pot, one-catalyst synthesis of graft copolymers by controlled ROMP and ATRP polymerizations.
Charvet R and Novak BM.
Macromolecules, 37(23), 8808-8811 (2004)
Feng Yan et al.
Chemical communications (Cambridge, England), (25)(25), 2696-2698 (2006-06-21)
Surfactants based on imidazolium ionic liquids (ILs), including polymerizable surfactant ILs, have been synthesized and used to stabilize polymerizable microemulsions useful for producing polymer nanoparticles, gels, and open-cell porous materials.
Tatiana N Pashirova et al.
ChemPlusChem, 85(8), 1939-1948 (2020-09-01)
The aim of this work was to increase the efficiency of catalytic systems for the hydrolytic cleavage of 4-nitrophenyl esters of phosphonic acids. Quaternary ammonium-containing comb-like polyelectrolytes («polymerized micelles») with ester cleavable fragments and a low aggregation threshold were used
Vibrational spectroscopic studies of the attachment chemistry for zirconium phosphonate multilayers at gold and germanium surfaces.
Frey BL, et al
Langmuir, 9(7), 1815-1820 (1993)
Qilu Zhang et al.
ACS applied materials & interfaces, 7(44), 24585-24591 (2015-10-22)
The controlled self-assembly of self-propelled Janus micromotors may give the micromotors some potential applications in many fields. In this work, we design a kind of SiO2-Pt Janus catalytic micromotor functionalized by spiropyran (SP) moieties on the surface of the SiO2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service