Skip to Content
Merck
All Photos(3)

Documents

152951

Sigma-Aldrich

1-Bromooctane

99%

Synonym(s):

Octyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7Br
CAS Number:
Molecular Weight:
193.12
Beilstein:
1733136
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.7 (vs air)

vapor pressure

0.34 mmHg ( 25 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

201 °C (lit.)

mp

−55 °C

density

1.118 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCBr

InChI

1S/C8H17Br/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3

InChI key

VMKOFRJSULQZRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromooctane was used as an extraction solvent in the determination of volatile organic compounds in water samples. It was also used in the synthesis of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Sun et al.
Biosensors & bioelectronics, 100, 28-34 (2017-08-30)
This work designed a three-component polymer for the preparation of polymer dots (Pdots). The polymer contained 9-(diphenylmethylene)-9H-fluorene (DPF), 9,9-dioctyl-9H-fluorene (DOF) and 1,1'-binaphthyl moieties, and was synthesized via Pd-catalyzed Suzuki reaction. It exhibited obvious yellow-colored aggregation-induced emission (AIE) for fluorescence enhancement
Fatma Yurt Lambrecht et al.
Chemical biology & drug design, 90(1), 141-146 (2017-01-10)
Imidazolium salts and derivatives have antitumor efficacy and toxic effects in different micro-organisms. In this study, an imidazolium bromide salt (NMI) was synthesized, and its antitumor potential was investigated by in vitro studies. Radiolabeling of synthesized NMI was carried out
Mei-I Leong et al.
Journal of separation science, 35(5-6), 688-694 (2012-01-25)
Volatile organic compounds (VOCs) are toxic compounds in the air, water and land. In the proposed method, ultrasound-assisted emulsification microextraction (USAEME) combined with gas chromatography-mass spectrometry (GC-MS) has been developed for the extraction and determination of eight VOCs in water
Ekaterina Kolobova et al.
Talanta, 188, 183-191 (2018-07-22)
The subject of this study is comparison of imidazolium based dynamic and covalent coatings of the quartz capillary wall on the example of catecholamines determination. A way of synthesis of covalent coatings was proposed. For the first time different type
Parmila Kumari et al.
Journal of biochemistry, 162(6), 395-401 (2017-12-01)
Genome profiling-based mutation assay (GPMA) is, to date, the only DNA sequence-based mutation assay that directly measures DNA alterations induced by mutagens. Here, the all-important congruence of mutagen assignment between DNA-based GPMA and the phenotype-based Ames test (the gold standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service