Skip to Content
Merck
All Photos(1)

Documents

464953

SAFC

Isoprene

≥99%

Synonym(s):

2-Methyl-1,3-butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHC(CH3)=CH2
CAS Number:
Molecular Weight:
68.12
Beilstein:
969158
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.35 (vs air)

vapor pressure

8.82 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

428 °F

contains

100-150 ppm p-tert-butylcatechol as inhibitor

expl. lim.

10 %

refractive index

n20/D 1.422 (lit.)

bp

34 °C (lit.)

mp

−146 °C (lit.)

density

0.681 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C=C

InChI

1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3

InChI key

RRHGJUQNOFWUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isoprene may be used in the synthesis of asymmetric polystyrene-block-polyisoprene (PS-b-PI) diblock copolymers via sequential anionic polymerization with styrene in the presence of sec-butyllithium as initiator.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 1 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-65.2 °F - closed cup

Flash Point(C)

-54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is it useful to ask why plants emit isoprene?
Thomas D Sharkey
Plant, cell & environment, 36(3), 517-520 (2012-11-14)
Thomas D Sharkey et al.
Annals of botany, 101(1), 5-18 (2007-10-09)
Some, but not all, plants emit isoprene. Emission of the related monoterpenes is more universal among plants, but the amount of isoprene emitted from plants dominates the biosphere-atmosphere hydrocarbon exchange. The emission of isoprene from plants affects atmospheric chemistry. Isoprene
Jianming Yang et al.
PloS one, 7(4), e33509-e33509 (2012-05-05)
The depleting petroleum reserve, increasingly severe energy crisis, and global climate change are reigniting enthusiasm for seeking sustainable technologies to replace petroleum as a source of fuel and chemicals. In this paper, the efficiency of the MVA pathway on isoprene
Tomohisa Kuzuyama et al.
Natural product reports, 20(2), 171-183 (2003-05-09)
This review covers the biosynthesis of the starter units of terpenoids, isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP) via the nonmevalonate pathway together with a new enzyme involved in the conversion of IPP and DMAPP, i.e type 2 IPP isomerase.
Morphology and elasticity of polystyrene-block-polyisoprene diblock copolymers in the melt.
Haenelt TG, et al.
Korea-Australia Rheology Journal, 26(3), 263-275 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service