111805
trans-1,3-Pentadiene
90%
Synonym(s):
trans-Piperylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH=CHCH=CH2
CAS Number:
Molecular Weight:
68.12
Beilstein:
1523659
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
6.56 psi ( 20 °C)
Quality Level
Assay
90%
refractive index
n20/D 1.430 (lit.)
bp
42 °C (lit.)
mp
−87 °C (lit.)
density
0.678 g/mL at 20 °C
0.683 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C\C=C\C=C
InChI
1S/C5H8/c1-3-5-4-2/h3-5H,1H2,2H3/b5-4+
InChI key
PMJHHCWVYXUKFD-SNAWJCMRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Trans-1,3-Pentadiene spectra is recorded in the 5000-17500 cm(-1) region with conventional and intracavity laser photoacoustic spectroscopy. It can be determined by the spectrophotometric method based on the Diels-Alder reaction. In this reaction, cisoid 1,3-dienes and tetracyanoethylene (TCNE) react with an aromatic-TCNE pi-complex.
Application
- Hydroxyl radical reactions with diolefins in atmospheric and combustion environments: This study provides insights into the reactions of hydroxyl radicals with diolefins such as trans-1,3-Pentadiene, highlighting their significance in both atmospheric chemistry and combustion processes. This research helps in understanding the reactive pathways and kinetics that could inform both environmental and industrial applications of trans-1,3-Pentadiene (Khaled et al., 2019).
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
5.0 °F - closed cup
Flash Point(C)
-15 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D A Williams et al.
Talanta, 20(11), 1085-1096 (1973-11-01)
An indirect spectrophotometric method, based on the rapid Diels-Alder reaction between cisoid 1,3-dienes and tetracyanoethylene (TCNE) and the destruction of an aromatic-TCNE pi-complex, was developed to determine eleven 1,3-dienes in the 0.05-1.00 x 10(-3)M range. These dienes were: cyclopentadiene; 1,3-cyclohexadiene;
Ruth J Waldrom et al.
The journal of physical chemistry. A, 110(3), 913-920 (2006-01-20)
The room-temperature vapor-phase overtone spectra of cis- and trans-1,3-pentadiene (piperylene) have been recorded in the 5000-17500 cm(-1) region with the use of conventional and intracavity laser photoacoustic spectroscopy. The presence of five nonequivalent olefinic CH bonds and one methyl group
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service