Skip to Content
Merck
All Photos(1)

Key Documents

295035

Sigma-Aldrich

1,3-Butadiene

≥99%

Synonym(s):

Bivinyl, Vinylethylene, alpha,gamma-Butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH=CH2
CAS Number:
Molecular Weight:
54.09
Beilstein:
605258
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

1.9 (15 °C, vs air)

Quality Level

vapor pressure

1863 mmHg ( 21 °C)

Assay

≥99%

autoignition temp.

788 °F

contains

p-tert-butylcatechol as inhibitor

expl. lim.

12 %

bp

−4.5 °C (lit.)

mp

−109 °C (lit.)

solubility

water: soluble 0.5 g/L at 20 °C

density

0.62 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC=C

InChI

1S/C4H6/c1-3-4-2/h3-4H,1-2H2

InChI key

KAKZBPTYRLMSJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Butadiene (BD) is a simplest conjugated diene. It is a colorless gas that turns to liquid easily. The vapor phase conversion of ethanol in the presence of Cr-Ba/MCM-41 (mesoporous silica molecular sieve) catalyst reported a yield of 25% of BD. It is also formed as a coproduct of ethylene production.[1] Mechanism of Diels-Alder reaction of 1,3-butadiene with ethylene has been investigated.[2]
May contain up to 4% dimer impurity.

Application

1,3-Butadiene can undergo a four-component coupling reaction with aryl Grignard reagents, and alkyl fluorides in the presence of nickel catalyst to form 1,6-octadiene carbon compound substituted with alkyl and aryl groups at the 3- and 8-positions.[3]
1,3-Butadiene is a useful diene for Diels Alder reaction.[4]
It may be used in the synthesis of the following:
  • 1-Silyl-substituted 1,3-butadienes, by [RuHCl(CO)(PCy3)2]-catalyzed silylative coupling of terminal (E)-1,3-dienes with vinylsilanes.[5]
  • Synthetic rubber and thermoplastic resins.[1]
  • Disilylated dimers by reacting with chlorosilanes.[6]
  • Octa-2,7-dien-1-ol via palladium catalyzed-hydrodimerization.[7]

Biochem/physiol Actions

1,3-Butadiene has been reported to induce hemangiosarcomas of the heart, malignant lymphomas, alveolar-bronchiolar neoplasms, squamous cell neoplasms of the forestomach in males and female mice. It also induced acinar cell carcinomas of the mammary gland, granulosa cell neoplasms of the ovary, and hepatocellular neoplasms in female mice.[5] Carcinogenicity induced by inhaled 1,3-butadiene has been investigated in C57BL/6 x C3H F1 mice.[8]
Environmental carcinogen. Induces cardiac hemangiosarcomas in mice.

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-104.8 °F - closed cup

Flash Point(C)

-76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-catalyzed coupling reaction of alkyl halides with aryl Grignard reagents in the presence of 1, 3-butadiene: Mechanistic studies of four-component coupling and competing cross-coupling reactions.
Iwasaki T, et al.
Chemical Science (2018)
J E Huff et al.
Science (New York, N.Y.), 227(4686), 548-549 (1985-02-01)
Groups of 50 male and 50 female B6C3F1 mice were exposed 6 hours per day, 5 days per week, for 60 to 61 weeks to air containing 0, 625, or 1250 parts per million 1,3-butadiene. These concentrations are somewhat below
Carcinogenicity of 1,3-butadiene in C57BL/6 x C3H F1 mice at low exposure concentrations.
Melnick RL, et al.
Cancer Research, 50, 6592- 6599 (1990)
Palladium-Catalyzed Dimerization Disilylation of 1,3-Butadiene with Chlorosilanes.
Terao J, et al.
Organic Letters, 6(19), 3341-3344 (2004)
Useful diene for the Diels-Alder reaction.
Danishefsky S and Kitahara T.
Journal of the American Chemical Society, 96(25), 7807-7808 (1974)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service