Skip to Content
Merck
All Photos(2)

Key Documents

459674

Sigma-Aldrich

Nickel(II) bromide 2-methoxyethyl ether complex

Synonym(s):

Diglyme nickel dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiBr2 · O(CH2CH2OCH3)2
CAS Number:
Molecular Weight:
352.67
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

Quality Level

reaction suitability

reagent type: catalyst
core: nickel

SMILES string

Br[Ni]Br.COCCOCCOC

InChI

1S/C6H14O3.2BrH.Ni/c1-7-3-5-9-6-4-8-2;;;/h3-6H2,1-2H3;2*1H;/q;;;+2/p-2

InChI key

RRSIMIHTHWYRRA-UHFFFAOYSA-L

Application

Nickel(II) bromide 2-methoxyethyl ether complex can be used as a catalyst to prepare:
  • Hindered diarylmethanols and benzylic alcohols by the addition of aryl halides to various aldehydes in the presence of Zn as a reducing agent.
  • Benzene-fused cyclic compounds via reductive dicarbofunctionalization of unactivated alkenes using less reactive benzyl chlorides.
  • Benzylboron derivatives via C-C cross-coupling reaction between substituted 5-bromo-3-methoxy-2-(trimethylsilyl)phenyl triflate and bromomethylboronic acid pinacol ester.
  • N-(Hetero)aryl sulfamides by N-arylation of sulfamides with aryl bromides in the presence of a photosensitizer.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-catalyzed asymmetric reductive arylalkylation of unactivated alkenes
Jin Youxiang and Wang Chuan
Angewandte Chemie (International Edition in English), 58(20), 6722-6726 (2019)
Nickel-Catalyzed Addition of Aryl Bromides to Aldehydes To Form Hindered Secondary Alcohols
Garcia KJ, et al.
Journal of the American Chemical Society, 141(5), 1823-1827 (2019)
Photochemically Mediated Nickel-Catalyzed Synthesis of N-(Hetero) aryl Sulfamides
Simons RT, et al.
The Journal of Organic Chemistry, 85(10), 6380-6391 (2020)
Aryne Reaction and Cross-coupling Approach for the Synthesis of Diverse N-Arylphenylalanine Derivatives
Kobayashi T, et al.
Chemistry Letters (Jpn), 49(7), 809-812 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service