Skip to Content
Merck
All Photos(3)

Documents

442712

Sigma-Aldrich

Lithium iron(III) oxide

greener alternative

95%

Synonym(s):

Battery_Electrode_Material_Cathode, Lithium ferrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiFeO2
CAS Number:
Molecular Weight:
94.78
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

mol wt

Mw 94.78 g/mol

composition

LiFeO2

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<1 μm

density

1.9 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[O-][Fe]=O

InChI

1S/Fe.Li.2O/q;+1;;-1

InChI key

JXGGISJJMPYXGJ-UHFFFAOYSA-N

General description

Lithium iron(III) oxide is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LiFeO2 can be used as a non-toxic cathode material with a specific capacity of ~120 mAhg−1 at a rate of 100 mAg−1. It can further be used in the fabrication of lithium-ion batteries and rechargeable batteries.
Studied for its use as a long-lived porous cathode in molten carbonate fuel cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and electrochemical properties of a new cathode material, LiFeO2, with a tunnel structure
Matsumura T, et al.
Journal of the Electrochemical Society, 149(12), A1509-A1513 (2002)
Synthesis of nano-crystalline LiFeO2 material with advanced battery performance
Lee YS, et al.
Electrochemical Communications, 4(9), 727-731 (2002)
Synthesis of tetrahedral LiFeO2 and its behavior as a cathode in rechargeable lithium batteries
Armstrong AR, et al.
Journal of the American Chemical Society, 130(11), 3554-3559 (2008)
Chitosan nanoparticles: a new vehicle for the improvement of the delivery of drugs to the ocular surface. Application to cyclosporin A
De Campos AM, et al.
International Journal of Pharmaceutics, 224(1-2), 159-168 (2001)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service