Skip to Content
Merck
All Photos(1)

Key Documents

751499

Sigma-Aldrich

Europium(II) iodide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Europium diiodide, Europium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
EuI2
CAS Number:
Molecular Weight:
405.77
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
form:
beads
Pricing and availability is not currently available.

grade

for synthesis

product line

AnhydroBeads

Assay

99.999% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

SMILES string

I[Eu]I

InChI

1S/Eu.2HI/h;2*1H/q+2;;/p-2

InChI key

DPYXWFUVSMSNNV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Europium Bromide is a ferromagnetic material[1] that can be used in the synthesis of bromide salt[2].

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanchez, J.P.; et al.
Inorganic Chemistry, 24, 408-415 (1985)
Gautam Gundiah et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 11), i76-i77 (2009-01-01)
Single crystals of Ba(0.96)Eu(0.04)BrI (barium europium bromide iodide) were grown by the Bridgman technique. The title compound adopts the ordered PbCl(2) structure [Braekken (1932 ▶). Z. Kristallogr.83, 222-282]. All atoms occupy the fourfold special positions (4c, site symmetry m) of
Cordier, Stephane; et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 619, 621-627 (1993)
Krings, M.;
Chemistry of Materials, 23, 1694-1694 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service