Skip to Content
Merck
All Photos(2)

Documents

425664

Sigma-Aldrich

Dysprosium(III) trifluoromethanesulfonate

98%

Synonym(s):

Dysprosium(III) triflate, Tris(triflato)dysprosium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Dy
CAS Number:
Molecular Weight:
609.71
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: dysprosium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

SMILES string

[Dy+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Dy/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

XSVCYDUEICANRJ-UHFFFAOYSA-K

General description

Dysprosium(III) trifluoromethanesulfonate (Dysprosium(III) triflate) is a lanthanide triflate.

Application

Catalyst for:
  • Aza-Piancatelli rearrangement
  • Friedel-Crafts alkylation
  • Ring-opening polymerization reactions
  • Microwave-assisted Kabachnik-Fields condensation
  • Cycloaddition reactions (Lewis-acid catalyst)
  • Fries rearrangement
  • Enantioselective glyoxalate-ene reactions
Dysprosium(III) trifluoromethanesulfonate, a water-tolerant Lewis acid, has been used in the following studies:
  • Aldol reaction of silyl enol ethers with aldehydes.
  • As an effective catalyst for electrophilic substitution reactions of indoles with imines.
  • As catalyst for the synthesis of 4-aminocyclopentenones and functionalized azaspirocycles, via intramolecular aza-Piancatelli rearrangement.
  • As new curing initiator to study the curing of diglycidyl ether of bisphenol-A (DGEBA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xie, W. et al.
Synlett, 498-498 (1999)
Study of lanthanide triflates as new curing initiators for DGEBA.
Castell P, et al.
Polymer, 41(24), 8465-8474 (2000)
Direct and highly diastereoselective synthesis of azaspirocycles by a dysprosium(III) triflate catalyzed aza-Piancatelli rearrangement.
Leoni I Palmer et al.
Angewandte Chemie (International ed. in English), 50(31), 7167-7170 (2011-06-21)
Versatile method for the synthesis of 4-aminocyclopentenones: dysprosium(III) triflate catalyzed aza-piancatelli rearrangement.
Gesine K Veits et al.
Angewandte Chemie (International ed. in English), 49(49), 9484-9487 (2010-11-06)
The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service