Skip to Content
Merck
All Photos(1)

Key Documents

402192

Sigma-Aldrich

3-Formyl-6-nitrochromone

99%

Synonym(s):

6-Nitro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5NO5
CAS Number:
Molecular Weight:
219.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

mp

157-161 °C (lit.)

functional group

aldehyde
ketone
nitro

SMILES string

[O-][N+](=O)c1ccc2OC=C(C=O)C(=O)c2c1

InChI

1S/C10H5NO5/c12-4-6-5-16-9-2-1-7(11(14)15)3-8(9)10(6)13/h1-5H

InChI key

JBDRQGWTNRIJRV-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

General description

3-Formyl-6-nitrochromone (6-Nitro-4-oxo-4H-1-benzopyran-3-carboxaldehyde) is a benzopyran derivative. It is a bicyclic heterocyclic molecule made up of a benzene ring fused to a heterocyclic pyran ring. 3-Formyl-6-nitrochromone has been reported to exhibit inhibitory activity against urease.[1]

Application

3-Formyl-6-nitrochromone is the suitable reagent used in a study to investigate the multidrug resistance reversal by some 3- formylchromones in human colon cancer and mouse lymphoma cells transfected with the human MDR1 gene.[2] It is the suitable reagent used in the synthesis of uridine-based library.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howard C Hang et al.
Chemistry & biology, 11(3), 337-345 (2004-05-05)
The polypeptide N-acetyl-alpha-galactosaminyltransferases (ppGalNAcTs, also abbreviated ppGaNTases) initiate mucin-type O-linked glycosylation and therefore play pivotal roles in cell-cell communication and protection of tissues. In order to develop new tools for studying mucin-type O-linked glycosylation, we screened a 1338 member uridine-based
Masami Kawase et al.
In vivo (Athens, Greece), 21(5), 829-834 (2007-11-21)
Several 3-formylchromone derivatives were examined for their tumor cell-cytotoxic, anti-Helicobacter pylori, urease inhibitory and anti-HIV activity. Comparing their relative cytotoxicity against four human tumor cell lines and three normal human cells, tumor cell-specific cytotoxicity was detected in some 3-formylchromone derivatives.
Zoltán Baráth et al.
In vivo (Athens, Greece), 20(5), 645-649 (2006-11-10)
Several new 3-formylchromone derivatives proved to be modifiers of multidrug resistance in mouse lymphoma cells and in human Colo320 colon cancer cells. There is apparently a structure-activity relationship between the antiproliferative multidrug resistance-reversing effect and the chemical structure of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service