Skip to Content
Merck
All Photos(1)

Key Documents

358665

Sigma-Aldrich

Tetrabutylammonium cyanide

95%

Synonym(s):

N,N,N-tributyl-1-butanaminium cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(CN)
CAS Number:
Molecular Weight:
268.48
Beilstein:
3598808
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

89-92 °C (lit.)

SMILES string

[C-]#N.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CN/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2/h5-16H2,1-4H3;/q+1;-1

InChI key

KRRBFUJMQBDDPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium cyanide (TBACN) can be used as a source of cyanide ion for colorimetric studies.
It can be employed as a reactant in the preparation of:
  • Tetrabutylammonium fluoride (TBAF), which can be used to remove silyl ether protecting groups.
  • Rhenium-based single-chain magnets, (DMF)4MReCl4(CN)2 (M = Mn, Fe, Co, Ni).
  • Phosphorescent blue light-emitting anionic iridium complexes.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, and DFT/TD-DFT calculations of highly phosphorescent blue light-emitting anionic iridium complexes
Di Censo D, et al.
Inorganic Chemistry, 47(3), 980-989 (2008)
Anhydrous tetrabutylammonium fluoride
Sun H and DiMagno SG
Journal of the American Chemical Society, 127(7), 2050-2051 (2005)
[ReCl4 (CN) 2] 2-: A High Magnetic Anisotropy Building Unit Giving Rise to the Single-Chain Magnets (DMF) 4MReCl4 (CN) 2 (M= Mn, Fe, Co, Ni)
Harris TD, et al.
Journal of the American Chemical Society, 132(11), 3980-3988 (2010)
Colorimetric detection of cyanide with a chromogenic oxazine
Tomasulo M and Raymo FM
Organic Letters, 7(21), 4633-4636 (2005)
A J Chinchalikar et al.
The European physical journal. E, Soft matter, 35(7), 55-55 (2012-07-06)
Clouding is studied by small-angle neutron scattering (SANS) on a charged micellar system of sodium dodecyl sulphate (SDS) and tetrabutylammonium bromide (TBAB) with varying temperature and salt NaCl. We show that the clouding occurs as a result of increase in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service