Skip to Content
Merck
All Photos(2)

Key Documents

399116

Sigma-Aldrich

Gallium(III) iodide

99.99% trace metals basis

Synonym(s):

Gallium triiodide, Triiodogallane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaI3
CAS Number:
Molecular Weight:
450.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤150.0 ppm Trace Metal Analysis

mp

345 °C (subl.) (lit.)

density

4.15 g/mL at 25 °C (lit.)

SMILES string

I[Ga](I)I

InChI

1S/Ga.3HI/h;3*1H/q+3;;;/p-3

InChI key

DWRNSCDYNYYYHT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gallium(III) iodide is a high melting pointinorganic solid with low volatility. It is used in various fields such as the production of semiconductors, gallium nitride crystals, and organic synthesis.

Application

Gallium(III) iodide can be used:
  • As a catalyst for Biginelli-type reaction to synthesize dihydropyrimidine-2(1H)-one derivatives.
  • As a reagent for the stereoselective synthesis of (Z)-β-iodo Baylis–Hillman adducts and for the aza-Prins cyclization.
  • As a precursor to synthesize highly crystalline gallium nitride crystals for energy conversion devices.
  • As a starting material to synthesize high-purity chalcogenide glasses.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gallium (III) Iodide-catalyzed Biginelli-type Reaction Under Solvent-free Conditions: Efficient Synthesis of Dihydropyrimidine-2 (1H)-one Derivatives
Dapeng Li, et al.
Chin. J. Chem., 28, 2025-2032 (2010)
Gallium iodide/iodine as a versatile reagent for the aza-Prins cyclization: an expeditious synthesis of 4-iodopiperidines
J.S. Yadav, et al
Tetrahedron Letters, 49, 3330-3334 (2008)
Rapid synthesis of crystalline gallium nitride from solid precursors at atmospheric pressure
Charles H Wallace, et al.
Chemistry of Materials, 11, 2299-2301 (1999)
Preparation of high-purity chalcogenide glasses containing gallium(III) sulfide
A.P. Velmuzhov, et al.},
Journal of Non-Crystalline Solids, 593, 121786-121786 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service