Skip to Content
Merck
All Photos(3)

Documents

203319

Sigma-Aldrich

Gallium

99.9995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga
CAS Number:
Molecular Weight:
69.72
MDL number:
UNSPSC Code:
12141715
eCl@ss:
38120701
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9995% trace metals basis

form

(metallic liquid or solid (ambient temp dependent))

resistivity

25.795 μΩ-cm, 30°C

bp

2403 °C (lit.)

mp

29.8 °C (lit.)

density

5.904 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Ga]

InChI

1S/Ga

InChI key

GYHNNYVSQQEPJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The wide liquid range of gallium was exploited recently when it was used as a solvent for the successful synthesis of ternary silicides, SmNiSi3 and YNiSi3, without incorporating Ga in their structure -- a promising new synthetic method.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Jakupec et al.
Metal ions in biological systems, 42, 425-462 (2004-06-23)
Gallium has been the second metal to show activity against malignant tumors in humans soon after the establishment of platinum drugs in routine clinical practice. It has the unique property of inhibiting tumor growth as a simple cation, mainly because
Andrew B Kelson et al.
Current opinion in pharmacology, 13(5), 707-716 (2013-07-24)
Microbes have evolved elaborate iron-acquisition systems to sequester iron from the host environment using siderophores and heme uptake systems. Gallium(III) is structurally similar to iron(III), except that it cannot be reduced under physiological conditions, therefore gallium has the potential to
Michael A Jakupec et al.
Current topics in medicinal chemistry, 4(15), 1575-1583 (2004-12-08)
The trivalent gallium cation is capable of inhibiting tumor growth, mainly because of its resemblance to ferric iron. It affects cellular acquisition of iron by binding to transferrin, and it interacts with the iron-dependent enzyme ribonucleotide reductase, resulting in reduced
Prasana Sahoo et al.
Biosensors & bioelectronics, 44, 164-170 (2013-02-22)
We demonstrate a very simple and generic protocol for ultrasensitive in-situ label-free detection of DNA hybridization using third generation poly(amidoamine)dendrimer (G3-PAMAM) functionalized GaN nanowires (NWs). PAMAM modified GaN NWs provides large density of docking site to immobilize significant number of
Christopher R Chitambar et al.
Antioxidants & redox signaling, 18(8), 956-972 (2012-08-21)
Despite advances made in the treatment of cancer, a significant number of patients succumb to this disease every year. Hence, there is a great need to develop new anticancer agents. Emerging data show that malignant cells have a greater requirement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service