Skip to Content
Merck
All Photos(1)

Key Documents

578606

Sigma-Aldrich

Indium(I) iodide

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Indium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InI
CAS Number:
Molecular Weight:
241.72
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: indium

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

[In]I

InChI

1S/HI.In/h1H;/q;+1/p-1

InChI key

FOVZCYAIUZHXGB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Selective recovery of indium from iron-rich solutions using an Aliquat 336 iodide supported ionic liquid phase (SILP): This study investigates the selective recovery of indium via adsorption by forming indium iodide complexes, demonstrating a preference for indium over iron (Van Roosendael et al., 2019).
  • Experimental evaluation of indium (I) iodide as a lead-free perovskite-inspired material for photovoltaic applications: This research explores the photovoltaic properties of indium(I) iodide thin films, suggesting its potential as an eco-friendly alternative in solar cell technologies (Ustinova et al., 2022).
  • Photovoltaic effect in Indium (I) iodide thin films: The study focuses on the deposition and photovoltaic effects of indium(I) iodide films, evaluating its viability for photovoltaic applications (Dunlap-Shohl et al., 2018).
  • Indium-mediated stereoselective allylation: This publication discusses the indium-mediated allylation processes, particularly the use of indium(I) iodide, highlighting its efficiency and selectivity in organic synthesis (Kumar et al., 2016).
  • Copper-catalyzed and indium-mediated methoxycarbonylation of unactivated alkyl iodides with balloon CO: The article covers the development of a method using indium(I) iodide for the methoxycarbonylation of alkyl iodides, providing a new pathway for ester formation (Chen et al., 2019).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service