Skip to Content
Merck
All Photos(3)

Documents

323403

Sigma-Aldrich

Lithium aluminum hydride

greener alternative

pellets, reagent grade, 95%

Synonym(s):

LAH, Lithium alanate, Lithium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

95%

form

pellets

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

size

10 mm × 6 mm

mp

125 °C (dec.) (lit.)

greener alternative category

SMILES string

[Li].[AlH3]

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium aluminum anhydride is effective as a selective reducing agent.The anhydride may be prepared by reacting lithium hydride with aluminum chloride. It is soluble in ether and it can reduce carboxyl group to primary alcohol group with great ease. Other organic compounds that can be reduced by lithium aluminum hydride are aldehydes, ketones, acid chlorides, acid anhydrides, halides, quinones, nitrogen compounds.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium aluminum hydride (LAH) can be used as a promising material for hydrogen storage applications. Its properties include high gravimetric and volumetric hydrogen densities.
Lithium aluminum hydride can be used to prepare undec-10-en-1-ol, a key intermediate for the preparation of polythioethers.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sasol Encyclopaedia of Science and Technology null
Renewable sulfur-containing thermoplastics via AB-type thiol-ene polyaddition
van den Berg O, et al.
European Polymer Journal, 49(4), 804-812 (2013)
Reduction of Organic Compounds by Lithium Aluminum Hydride. III. Halides, Quinones, Miscellaneous Nitrogen Compounds1.
Nystrom RF and Brown WG
Journal of the American Chemical Society, 70(11), 3738-3740 (1948)
Reversible hydrogen storage via titanium-catalyzed LiAlH4 and Li3AlH6
Chen J, et al.
The Journal of Physical Chemistry B, 105(45), 11214-11220 (2001)
Reduction of organic compounds by lithium aluminum hydride. I. Aldehydes, ketones, esters, acid chlorides and acid anhydrides.
Nystrom RF and Brown WG
Journal of the American Chemical Society, 69(5), 1197-1199 (1947)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service