Skip to Content
Merck
Lithium aluminum hydride ≥97.0% (gas-volumetric)

Lithium aluminum hydride

Lithium aluminum hydride ≥97.0% (gas-volumetric)
Synonym(s):
LAH, Lithium alanate, Lithium tetrahydroaluminate
Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade