Skip to Content
Merck
All Photos(1)

Key Documents

303453

Sigma-Aldrich

3-Bromo-2-coumaranone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5BrO2
CAS Number:
Molecular Weight:
213.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

mp

107-109 °C (lit.)

solubility

ethyl acetate: soluble 25 mg/mL, clear, colorless to yellow

functional group

bromo

storage temp.

2-8°C

SMILES string

BrC1C(=O)Oc2ccccc12

InChI

1S/C8H5BrO2/c9-7-5-3-1-2-4-6(5)11-8(7)10/h1-4,7H

InChI key

MXSVEBOOHMKMQG-UHFFFAOYSA-N

General description

3-Bromo-2-coumaranone has been tested for allergenicity by murine local lymph-node assay.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Hostýnek et al.
Archives of dermatological research, 287(6), 567-571 (1995-01-01)
A structure-activity relationship model for prediction of contact allergenic potential of chemicals had previously been developed. The model had been shown to be able to classify known allergens and nonallergens using data on physicochemical and reactivity parameters of functional groups

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service