Skip to Content
Merck
All Photos(3)

Key Documents

E44205

Sigma-Aldrich

4-Ethylphenol

99%

Synonym(s):

(4-Hydroxyphenyl)ethane, (p-Hydroxyphenyl)ethane, 1-Ethyl-4-hydroxybenzene, 4-Ethylphenol, p-Ethylphenol, p-Hydroxyethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1363317
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023356
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.2 (vs air)

vapor pressure

0.13 mmHg ( 20 °C)

Assay

99%

bp

218-219 °C (lit.)

mp

40-42 °C (lit.)

SMILES string

CCc1ccc(O)cc1

InChI

1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3

InChI key

HXDOZKJGKXYMEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyrille Sabot et al.
Chemical communications (Cambridge, England), (20)(20), 2941-2943 (2009-05-14)
A concise total synthesis of (+/-)-aspidospermidine has been achieved in twelve steps from inexpensive 4-ethylphenol, based on an oxidative Hosomi-Sakurai reaction via the concept of "aromatic ring umpolung".
Hajo Zech et al.
Proteomics, 11(16), 3380-3389 (2011-07-14)
Two-dimensional difference gel electrophoresis (2-D DIGE) allows for reliable quantification of global protein abundance changes. The threshold of significance for protein abundance changes depends on the experimental variation (biological and technical). This study estimates biological, technical and total variation inherent
Michael R L Stratford et al.
Bioorganic & medicinal chemistry, 20(14), 4364-4370 (2012-06-16)
In vitro studies, using combined spectrophotometry and oximetry together with hplc/ms examination of the products of tyrosinase action demonstrate that hydroquinone is not a primary substrate for the enzyme but is vicariously oxidised by a redox exchange mechanism in the
Isa Silva et al.
International journal of food microbiology, 145(2-3), 471-475 (2011-02-19)
This work aimed to evaluate the effect of certain factors on the production of volatile phenols from the metabolism of p-coumaric acid by lactic acid bacteria (LAB) (Lactobacillus plantarum, L. collinoides and Pediococcus pentosaceus). The studied factors were: pH, L-malic
S Benito et al.
International journal of food microbiology, 132(2-3), 145-152 (2009-05-15)
Different strains of Saccharomyces with different hydroxycinnamate decarboxylase (HCDC) activities, estimated by a bioconversion assay, were used for the fermentation of musts enriched with p-coumaric acid and grape anthocyanins, with the aim of favouring the formation of vinylphenolic pyranoanthocyanins, colour

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service