All Photos(1)
About This Item
Linear Formula:
(CH3)2C=CH2
CAS Number:
Molecular Weight:
56.11
Beilstein:
773645
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
2 (vs air)
Quality Level
vapor pressure
3278 mmHg ( 37.7 °C)
Assay
99%
autoignition temp.
869 °F
bp
−6.9 °C (lit.)
mp
−140 °C
SMILES string
CC(C)=C
InChI
1S/C4H8/c1-4(2)3/h1H2,2-3H3
InChI key
VQTUBCCKSQIDNK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2-methylpropene is used to synthesize S-(+)-2,2-Dimethylcyclopropane carboxylic acid via asymmetric cyclopropanation reaction.
Application
2-Methylpropene can be used in the following reactions:
- Asymmetric synthesis of S-(+)-2,2-dimethylcyclopropane carboxylic acid, a key building block for the synthesis of cilastatin.
- Alkylation of p-xylene to form t-butyl-p-xylene catalyzed by a heteropolyacid.
- Synthesis of poly(2-methylpropene) or polyisobutene.
Packaging
Supplied in a Sure/Pac™ cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.
Compatible with the following:
Compatible with the following:
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
also commonly purchased with this product
Product No.
Description
Pricing
hose barb
Product No.
Description
Pricing
recommended
Product No.
Description
Pricing
regulator
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1 - Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
-112.0 °F - closed cup
Flash Point(C)
-80 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Selective alkylation of p-xylene with 2-methylpropene by 12-tungstophosphoric acid.
Soeda H, et al.
Chemistry Letters (Jpn), 23(5), 909-912 (1994)
The synthesis of S-(+)-2, 2-dimethylcyclopropane carboxylic acid: a precursor for cilastatin.
Wang Q, et al.
Tetrahedron Asymmetry, 9(22), 3971-3977 (1998)
The synthesis of S-(+)-2, 2-dimethylcyclopropane carboxylic acid: a precursor for cilastatin
Wang Q, et al.
Tetrahedron Asymmetry, 9, 3971-3977 (1998)
Molybdenum (III) Compounds as Catalysts for 2?Methylpropene Polymerization.
Hijazi A K, et al.
Angewandte Chemie (International Edition in English), 46(38), 7290-7292 (2007)
Solvent?Ligated Copper (II) Complexes for the Homopolymerization of 2?Methylpropene.
Li Y, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 14(26), 7997-8003 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service