Skip to Content
Merck
All Photos(1)

Key Documents

86262

Sigma-Aldrich

2-Methyl-2-butene

≥95.0% (GC)

Synonym(s):

β-Isoamylene, Amylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)2
CAS Number:
Molecular Weight:
70.13
Beilstein:
1361353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.4 (vs air)

Assay

≥95.0% (GC)

form

liquid

expl. lim.

8.7 %

refractive index

n20/D 1.385 (lit.)
n20/D 1.388

bp

35-38 °C (lit.)

mp

−134 °C (lit.)

density

0.662 g/mL at 25 °C (lit.)

SMILES string

C\C=C(\C)C

InChI

1S/C5H10/c1-4-5(2)3/h4H,1-3H3

InChI key

BKOOMYPCSUNDGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Methyl-2-butene is a trisubstituted olefin. It acts as guest and forms stable solid host-guest complexes with self-assembled benzophenone bis-urea macrocycles. The impact of active chlorine on photo-oxidation of 2-methyl-2-butene was studied. Photosensitized oxidation of 2-methyl-2-butene adsorbed on internal framework of Na-ZSM-5 zeolite was studied. Gas-phase reaction of 2-methyl-2-butene with ozone has been investigated. Kinetics of liquid-phase alkylation of 3-methylthiophene with 2-methyl-2-butene on supported phosphoric acid has been reported.

2-methyl-2-butene undergoes ultraviolet irradiation, leading to the formation of trimethylene oxides.

Application

2-Methyl-2-butene may be employed as precursor for peroxyacetyl nitrate for the calibration purposes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of liquid-phase alkylation of 3-methylthiophene with 2-methyl-2-butene over a solid phosphoric acid.
Belliere V, et al.
Applied Catalysis. B, Environmental, 64(3), 254-261 (2006)
Light-catalyzed organic reactions. I. The reaction of carbonyl compounds with 2-methyl-2-butene in the presence of ultraviolet light
Buchi G, et al.
Journal of the American Chemical Society, 76, 4327-4331 (1954)
Mehdi Nabi et al.
Journal of AOAC International, 103(5), 1243-1249 (2020-11-27)
A simple, rapid, selective and sensitive sample preparation and derivatization method was performed for determination of bromate ions in water by means of dispersive liquid-liquid extraction (DLLE) by gas chromatography-electron capture detection (GC-ECD). This method is based on 2-methyl-2-butene derivatization
Gas-phase reaction of ozone with 2-methyl-2-butene: Dicarbonyl formation from Criegee biradicals.
Grosjean D.
Environmental Science & Technology, 24(9), 1428-1432 (1990)
Light-catalyzed organic reactions. I. The reaction of carbonyl compounds with 2-methyl-2-butene in the presence of ultraviolet light
Buchi G, et al.
Journal of the American Chemical Society, 76, 4327-4331 (1954)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service