Skip to Content
Merck
All Photos(3)

Key Documents

186171

Sigma-Aldrich

n-Butyllithium solution

1.6 M in hexanes

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
1209227
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.6 M in hexanes

density

0.68 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Copolymerization of butadiene and styrene under the influence of n-butyllithium: This study explores the copolymerization process using n-butyllithium as a catalyst, which is significant for the development of new synthetic rubber materials (V Bronskaya et al., 2023).
  • Modifiers of n-Butyllithium in the Synthesis of Polybutadiene and Styrene-Butadiene Rubbers: Explores the use of n-butyllithium with different modifiers in the synthesis of industrial rubbers, important for understanding the control of polymer properties (VS Glukhovskoi et al., 2014).
  • Aggregation and Solvation of n-Butyllithium: Examines the aggregation behavior and solvation properties of n-butyllithium in various solvents, providing insights into its chemical interactions essential for organic synthesis (O Tai et al., 2017).

Packaging

The Kilo-Lab® cylinders are only available in the US. The cylinder types will vary according to country.

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Other Notes

May contain trace amounts of LiCl.

Legal Information

Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 111, 2981-2981 (1989)
Jennifer L Rutherford et al.
Journal of the American Chemical Society, 124(2), 264-271 (2002-01-10)
6Li and (13)C NMR spectroscopic studies were carried out on [(6)Li]n-BuLi and [(6)Li]PhLi (RLi) in toluene-d(8) containing the following diamines: N,N,N',N'-tetramethylethylenediamine (TMEDA), N,N,N',N'-tetraethylethylenediamine, 1,2-dipyrrolidinoethane, 1,2-dipiperidinoethane, N,N,N',N'-tetramethylpropanediamine, trans-(R,R)-N,N,N',N'-tetramethylcyclohexanediamine, and (-)-sparteine. Dimers of general structure (RLi)(2)S(2) (S = chelating diamine) are formed
Enantioselective total synthesis of the marine toxin (-)-gymnodimine employing a Barbier-type macrocyclization.
Ke Kong et al.
Angewandte Chemie (International ed. in English), 48(40), 7402-7405 (2009-09-04)
Deyu Li et al.
The Journal of organic chemistry, 73(6), 2373-2381 (2008-02-26)
A system consisting of a chiral lithium amide and n-BuLi in tol-d(8) solution was investigated with (1)H and (13)C INEPT DOSY, (6)Li and (15)N NMR, and other 2D NMR techniques. A mixed 2:1 trimeric complex was identified as the major
Michael A Tarselli et al.
Organic letters, 11(20), 4596-4599 (2009-10-09)
A procedure for the coupling of aliphatic imines with allylic and allenic alkoxides is described. The success of these studies was enabled by a unique reactivity profile of Ti(IV) isopropoxide/n-BuLi compared to well-known Ti(IV) isopropoxide/RMgX systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service