Skip to Content
Merck
All Photos(3)

Key Documents

255777

Sigma-Aldrich

Magnesium chloride hexahydrate

99.995% trace metals basis

Synonym(s):

Dichloromagnesium hexahydrate, Magnesium dichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline
solubility:
ethanol: slightly soluble(lit.)

grade

for analytical purposes

Assay

99.995% trace metals basis

form

crystalline

impurities

≤55.0 ppm Trace Metal Analysis

solubility

ethanol: slightly soluble(lit.)

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Magnesium chloride hexahydrate is a highly water soluble crystalline solid widely used in nanomaterial synthesis and thermal energy storage.

Application

Magnesium chloride hexahydrate can be used:
  • As a precursor to synthesize magnesium hydroxide nanoparticles for flame retardant applications.
  • As magnesium source to prepare hydroxyapatite nanoparticles for biomedical application.
  • To fabricate phase change materials for passive thermal control of lithium-ion batteries.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium incorporated hydroxyapatite nanoparticles: Preparation, characterization, antibacterial and larvicidal activity
B. Gayathri, et al.
Arabian Journal of Chemistry, 11, 645-654 (2018)
Dongmin An, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 356, 28-31 (2010)
Development of the inorganic composite phase change materials for passive thermal management of Li-ion batteries: material characterization
Yana Galazutdinova, et al.},
International Journal of Energy Research, 44, 2011-2022 (2020)
Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve
Xingchu Gong et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 93, 260-266 (2015-04-25)
Brittleness is one of the important material properties that influences the success or failure of powder compaction. We have discovered that the reciprocal of diametrical elastic strain at fracture is the most suitable tablet brittleness indices (TBIs) for quantifying brittleness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service