Skip to Content
Merck
All Photos(2)

Key Documents

M1028

Sigma-Aldrich

Magnesium chloride solution

for molecular biology, 1.00 M±0.01 M

Synonym(s):

MgCl2, dichloromagnesium, magnesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.52
grade:
Molecular Biology
for molecular biology
form:
liquid
solubility:
water: soluble

grade

Molecular Biology
for molecular biology

Quality Level

sterility

non-sterile; 0.2 μm filtered

form

liquid

concentration

1.00 M±0.01 M

technique(s)

DNA sequencing: suitable

impurities

DNAse, none detected
NICKase, none detected
Protease, none detected
RNAse, none detected

solubility

water: soluble

density

1.080 g/cm3

cation traces

heavy metals (as Pb): ≤5 ppm

suitability

suitable for electrophoresis
suitable for molecular biology

application(s)

cell analysis

foreign activity

DNase, RNase, NICKase and protease, none detected

SMILES string

Cl[Mg]Cl

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Magnesium chloride is a common ionic halide that readily dissolves in water. It plays various crucial biological roles in enzymology, cell membrane/wall structure, muscle cell physiology, and nucleic acid composition. Additionally, magnesium chloride serves as an essential co-factor in numerous enzymes, including DNAse, specific restriction enzymes, and Ribonuclease H. It is widely utilized to provide magnesium ions in diverse molecular biology applications, including PCR reactions.

Application

Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It has been used -
  • to prepare buffer to lyse cells for immunoprecipitation
  • as a supplement in Krebs-Henseleit buffer in chronic high-fat diet studies of aged mice.
  • in embedding 3-dimensional mtgase hydrogels in invitro studies of osteocyte differentitaion and interconnected cellular networks.
  • It has been used to protect the liver from oxidative damage.
  • to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

Features and Benefits

  • Available as a Ready-to-use solution which reduces the need for preparation time.
  • Heavy metals (as Pb): ≤5 ppm
  • Free from DNase, RNase, NICKase and protease

Components

Magnesium chloride solution contains 1M magnesium chloride in 18 megaohm water.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective Effects of Magnesium Chloride on Liver Enzymes and Biomarkers of Oxidative Stress in high fat diet fed Rats
Mohammed et, al
Nigerian Journal of Physiological Sciences : Official Publication of the Physiological Society of Nigeria, 34(2), 149-157 (2019)
Yelena Y Grinberg et al.
Journal of neurochemistry, 122(1), 221-229 (2012-04-25)
Spreading depression (SD), the likely cause of migraine aura and perhaps migraine, is triggered by widespread and unfettered neuronal hyperexcitability. Migraine and the initiating hyperexcitability of seizure, which involve oxidative stress (OS), are likely interrelated. Environmental enrichment (EE) decreases seizure
Antimo Migliaccio et al.
Methods in molecular biology (Clifton, N.J.), 776, 361-370 (2011-07-29)
Much evidence indicates that, with few exceptions, non-genomic actions of steroids are mediated by receptors universally known as nuclear receptors. Steroid receptors do not exhibit intrinsic tyrosine kinase activity. Nevertheless, they stimulate different signaling pathways in cytoplasm of target cells
M J Mc Garrigle et al.
European cells & materials, 31, 323-340 (2016-05-25)
Extracellular matrix (ECM) stiffness and cell density can regulate osteoblast differentiation in two dimensional environments. However, it is not yet known how osteoblast-osteocyte differentiation is regulated within a 3D ECM environment, akin to that existing in vivo. In this study
Hiroaki Eshima et al.
Journal of applied physiology (Bethesda, Md. : 1985), 128(5), 1153-1162 (2020-03-28)
Obesity and aging reduce skeletal muscle contractile function. However, it remains unclear whether obesity additively promotes muscle contractile dysfunction in the setting of aging. In this study, we investigated skeletal muscle contractile function ex vivo and intracellular Ca2+ release in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service