Skip to Content
Merck
All Photos(3)

Key Documents

217883

Sigma-Aldrich

Iron(III) bromide

greener alternative

98%

Synonym(s):

Ferric bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeBr3
CAS Number:
Molecular Weight:
295.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder

Quality Level

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

battery manufacturing

greener alternative category

SMILES string

Br[Fe](Br)Br

InChI

1S/3BrH.Fe/h3*1H;/q;;;+3/p-3

InChI key

FEONEKOZSGPOFN-UHFFFAOYSA-K

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service