400831
Iron(II) bromide
98%
Synonym(s):
Dibromoiron, Iron dibromide, Ferrous bromide
About This Item
Assay
98%
reaction suitability
core: iron
reagent type: catalyst
reaction type: Atom Transfer Radical Polymerization (ATRP)
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
bp
934 °C (lit.)
mp
684 °C (lit.)
density
4.63 g/mL at 25 °C (lit.)
greener alternative category
SMILES string
Br[Fe]Br
InChI
1S/2BrH.Fe/h2*1H;/q;;+2/p-2
InChI key
GYCHYNMREWYSKH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service