Skip to Content
Merck
All Photos(1)

Key Documents

8.03945

Sigma-Aldrich

Iron(III) chloride

greener alternative

anhydrous for synthesis

Synonym(s):

Iron(III) chloride, Ferric chloride, Iron trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl3Fe
CAS Number:
Molecular Weight:
162.20
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-729-4
NACRES:
NA.22
form:
powder
solubility:
920 g/L (Hydrolysis)

vapor pressure

1 hPa ( 20 °C)

Quality Level

form

powder

potency

316 mg/kg LD50, oral (Rat)

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

1 (20 °C, 200 g/L in H2O)

mp

306 °C (decomposition)

solubility

920 g/L (Hydrolysis)

density

2.89 g/cm3 at 25 °C

bulk density

1000 kg/m3

greener alternative category

storage temp.

2-30°C

SMILES string

[Fe](Cl)(Cl)Cl

InChI

1S/3ClH.Fe/h3*1H;/q;;;+3/p-3

InChI key

RBTARNINKXHZNM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Iron(III) chloride (FeCl3) can be used as a catalyst:
  • For the oxidation of glucose to gluconic acid in water.
  • To synthesize 2-iodo substituted benzo[b]thiophenes by iodocyclization of 2-alkynylthioanisoles in the presence of sodium iodide and ethanol as a solvent.
  • In the glycosylation sugars in the presence of allyl and alkynyl alcohols to synthesize corresponding glycosides.
  • To prepare 1,2-trans glycosyl azides by azido glycosylation of glycosyl β-peracetates.

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (Fe): passes test
Identity (Cl): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh B Salunke et al.
Chemical communications (Cambridge, England), 47(37), 10440-10442 (2011-08-16)
A highly efficient and mild method for azido glycosylation of glycosyl β-peracetates to 1,2-trans glycosyl azides was developed by using inexpensive FeCl(3) as the catalyst. In addition, we demonstrated, for the first time, that FeCl(3) in combination with copper powder
Iron (III) chloride catalyzed glycosylation of peracylated sugars with allyl/alkynyl alcohols
Narayanaperumal S, et al.
Journal of the Brazilian Chemical Society, 23, 1982-1988 (2012)
Green synthesis of benzo [b] thiophenes via iron (III) mediated 5-endo-dig iodocyclization of 2-alkynylthioanisoles
Kesharwani T, et al.
Tetrahedron Letters, 57(3), 411-414 (2016)
Oxidative conversion of glucose to gluconic acid by iron (III) chloride in water under mild conditions
Zhang H, et al.
Green Chemistry, 18(8), 2308-2312 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service