Skip to Content
Merck
All Photos(2)

Key Documents

203947

Sigma-Aldrich

Ammonium tetrachloropalladate(II)

≥99.995% trace metals basis

Synonym(s):

Palladium(II)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2PdCl4
CAS Number:
Molecular Weight:
284.31
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.995% trace metals basis
form:
solid

Quality Level

Assay

≥99.995% trace metals basis

form

solid

impurities

≤50.0 ppm Trace Metal Analysis

density

2.17 g/mL at 25 °C (lit.)

SMILES string

[H][N+]([H])([H])[H].[H][N+]([H])([H])[H].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.2H3N.Pd/h4*1H;2*1H3;/q;;;;;;+2/p-2

InChI key

IRKVTUVFHGUMMN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reacts with creatinine (cat. no. 859702) to form various Pd-creatinine complexes with applications in semiconductors, catalysis, mixed oxidation state studies, and electron conductivity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You Yu et al.
Small (Weinheim an der Bergstrasse, Germany), 11(28), 3444-3449 (2015-03-20)
Stretchable and transparent electrodes are fabricated by chemical deposition of metal thin films on natural veins of leaves at ambient conditions. These vein-based transparent electrodes show excellent electro-optical property (0.9 Ω sq(-1) at 83% T) even at 50% tensile strains
J Muris et al.
Allergy, 64(8), 1152-1156 (2009-02-12)
For patch testing, replacement of the commonly used palladium dichloride (PdCl2) by sodium tetrachloropalladate (Na2[PdCl4]) was recently demonstrated to improve test accuracy and show a significant correlation with nickel (Ni), supporting the concept of cross-reactivity between Pd and Ni. A
T N Semenets et al.
Radiatsionnaia biologiia, radioecologiia, 37(6), 892-895 (1998-02-19)
Efasol and potassium tetrachloropalladate being injected into mice prior to irradiation with a dose of 150 sGy increase the colony forming activity of animals bone marrow cells transplanted in lethally irradiated recipient mice. The palladium presence in such compounds is
G Pneumatikakis et al.
Journal of inorganic biochemistry, 37(1), 17-28 (1989-09-01)
The interactions of potassium tetrachloropalladate(II) with the B6 vitamins pyridoxal, pyridoxine, and pyridoxamine in 1:1 molar ratio have been studied. From DMF solutions, the ionic trichloro (pyridoxal or pyridoxine) palladates(II) were isolated. Pyridoxamine, on the other hand, in aqueous solutions
[Spectrophotometric analysis of D-penicillamine and mercaptopropionylglycine in pharmaceutical formulations with ammonium tetrachloropalladate].
M A Raggi et al.
Bollettino chimico farmaceutico, 125(8), 295-297 (1986-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service