Skip to Content
Merck
All Photos(2)

Key Documents

379808

Sigma-Aldrich

Sodium tetrachloropalladate(II)

≥99.99% trace metals basis

Synonym(s):

Palladium(II) sodium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2PdCl4
CAS Number:
Molecular Weight:
294.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
powder and chunks

Quality Level

Assay

≥99.99% trace metals basis

form

powder and chunks

impurities

≤100.0 ppm Trace Metal Analysis

SMILES string

[Na+].[Na+].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.2Na.Pd/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

ZIXLZKBNIAXVBE-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang Zhou et al.
Nature communications, 11(1), 335-335 (2020-01-19)
The sintering of supported metal nanoparticles is a major route to the deactivation of industrial heterogeneous catalysts, which largely increase the cost and decrease the productivity. Here, we discover that supported palladium/gold/platinum nanoparticles distributed at the interface of oxide supports
Ming Zhao et al.
ACS nano, 13(6), 7241-7251 (2019-05-31)
Owing to their highly open structure and a large number of low-coordination sites on the surface, noble-metal nanoframes are intriguing for catalytic applications. Here, we demonstrate the rational synthesis of Ru cuboctahedral nanoframes with enhanced catalytic performance toward hydrazine decomposition.
Charles L Gaworski et al.
Inhalation toxicology, 20(2), 167-182 (2008-02-01)
The use of a palladium (Pd) catalyst has been proposed to promote combustion of tobacco, thereby reducing concentrations of certain toxic components of smoke, including polyaromatic hydrocarbons (PAHs). In the present work, toxicological comparisons were made using experimental cigarettes containing
J Muris et al.
Allergy, 64(8), 1152-1156 (2009-02-12)
For patch testing, replacement of the commonly used palladium dichloride (PdCl2) by sodium tetrachloropalladate (Na2[PdCl4]) was recently demonstrated to improve test accuracy and show a significant correlation with nickel (Ni), supporting the concept of cross-reactivity between Pd and Ni. A
T N Semenets et al.
Radiatsionnaia biologiia, radioecologiia, 37(6), 892-895 (1998-02-19)
Efasol and potassium tetrachloropalladate being injected into mice prior to irradiation with a dose of 150 sGy increase the colony forming activity of animals bone marrow cells transplanted in lethally irradiated recipient mice. The palladium presence in such compounds is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service