Skip to Content
Merck
All Photos(3)

Key Documents

205818

Sigma-Aldrich

Sodium tetrachloropalladate(II)

98%

Synonym(s):

Palladium(II) sodium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2PdCl4
CAS Number:
Molecular Weight:
294.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder and chunks

Quality Level

Assay

98%

form

powder and chunks

SMILES string

[Na+].[Na+].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.2Na.Pd/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

ZIXLZKBNIAXVBE-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Sodium tetrachloropalladate(II)[Na2PdCl4] was used to prepared palladium nanospheres, hollow nanopheres and hollow AuxPdY alloy nanoparticles. Na2PdCl4 maybe used in the electroless coating on mild steel and to prepare ion associate cetyltrimethylammonium-tetrachloropalladate which is consequently used to modify carbon based electrodes for better sensitivity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenni Wang et al.
ACS nano, 11(1), 163-170 (2017-01-25)
Synthesis of metal nanoframes has been of great interest for their open structures and high fractions of active surface sites, which gives rise to outstanding performance in catalysis. In this work, Pd nanoframes with well-defined structures have been successfully prepared
Hollow flower-like AuPd alloy nanoparticles: One step synthesis, self-assembly on ionic liquid-functionalized graphene, and electrooxidation of formic acid
Chai J, et al.
Journal of Materials Chemistry, 21(44), 17922-17929 (2011)
Yaming Liu et al.
Nanoscale, 10(16), 7505-7510 (2018-04-12)
To date, great progress has been made in the shape-controlled synthesis of noble-metal nanocrystals. However, there still exists a major gap between academic studies and industrial applications due to the inability to produce nanocrystals in large quantities while retaining their
Charles L Gaworski et al.
Inhalation toxicology, 20(2), 167-182 (2008-02-01)
The use of a palladium (Pd) catalyst has been proposed to promote combustion of tobacco, thereby reducing concentrations of certain toxic components of smoke, including polyaromatic hydrocarbons (PAHs). In the present work, toxicological comparisons were made using experimental cigarettes containing
J Muris et al.
Allergy, 64(8), 1152-1156 (2009-02-12)
For patch testing, replacement of the commonly used palladium dichloride (PdCl2) by sodium tetrachloropalladate (Na2[PdCl4]) was recently demonstrated to improve test accuracy and show a significant correlation with nickel (Ni), supporting the concept of cross-reactivity between Pd and Ni. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service