Skip to Content
Merck
All Photos(1)

Documents

202134

Sigma-Aldrich

Cesium iodide

99.9% trace metals basis

Synonym(s):

Caesium iodide, Caesium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsI
CAS Number:
Molecular Weight:
259.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

impurities

≤1500.0 ppm Trace Metal Analysis

mp

626 °C (lit.)

density

4.51 g/mL at 25 °C (lit.)

SMILES string

[I-].[Cs+]

InChI

1S/Cs.HI/h;1H/q+1;/p-1

InChI key

XQPRBTXUXXVTKB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cesium iodide (CsI) can be used as a cathode for the fabrication of field emission cathodes for potential usage in high power microwave systems. It can also be used to form an interface layer between the blocking layer and the perovskite active layer to improve the photovoltaic performance and stability of solar cells.

Features and Benefits

Frequently used in devices such as phosphor screens for medical imaging, scintillators, calorimeters and a variety of particle detectors.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low level plasma formation in a carbon velvet cesium iodide coated cathode
Shiffler D, et al.
Physics of Plasmas, 11(4), 1680-1684 (2004)
Comparison of velvet-and cesium iodide-coated carbon fiber cathodes
Shiffler D, et al.
IEEE transactions on nuclear science, 29(3), 445-451 (2001)
Cong Chen et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(11), 1802046-1802046 (2019-06-11)
Photovoltaic devices employing lead halide perovskites as the photoactive layer have attracted enormous attention due to their commercialization potential. Yet, there exists challenges on the way to the practical use of perovskite solar cells (PSCs), such as light stability and
Olga Nazarenko et al.
Inorganic chemistry, 56(19), 11552-11564 (2017-09-13)
Interest in hybrid organic-inorganic lead halide compounds with perovskite-like two-dimensional crystal structures is growing due to the unique electronic and optoelectronic properties of these compounds. Herein, we demonstrate the synthesis, thermal and optical properties, and calculations of the electronic band
Marco Bertolini et al.
Medical physics, 39(5), 2617-2627 (2012-05-09)
The purpose of this study is to compare digital radiography systems using the metric effective detective quantum efficiency (eDQE), which better reflects digital radiography imaging system performance under clinical operating conditions, in comparison with conventional metrics such as modulation transfer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service