Skip to Content
Merck
All Photos(2)

Key Documents

203033

Sigma-Aldrich

Cesium iodide

99.999% trace metals basis

Synonym(s):

Caesium iodide, Caesium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsI
CAS Number:
Molecular Weight:
259.81
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38100503
PubChem Substance ID:
NACRES:
NA.23
form:
solid

Quality Level

Assay

99.999% trace metals basis

form

solid

impurities

≤15.0 ppm Trace Metal Analysis

mp

626 °C (lit.)

density

4.51 g/mL at 25 °C (lit.)

SMILES string

[I-].[Cs+]

InChI

1S/Cs.HI/h;1H/q+1;/p-1

InChI key

XQPRBTXUXXVTKB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium iodide can be used as precursor to synthesize lead-free perovskite material, Cs2NaBiI6 (CNBI). The CNBI is highly stable and finds application in the field of solar cells, LEDs, and lasers.

It can be used to prepare brightest red emitting Cs2HfI6 scintillator which is applicable in high resolution gamma spectroscopy.

It can also be used tosynthesize Cesium based nanocrystals for the detection of ionizingradiations.

Features and Benefits

  • High quantum efficiency
  • High stability to ambient air and gas environment
Frequently used in devices such as phosphor screens for medical imaging, scintillators, calorimeters and a variety of particle detectors.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Hajdok et al.
Medical physics, 35(7), 3194-3204 (2008-08-14)
A frequency-dependent x-ray Swank factor based on the "x-ray interaction" modulation transfer function and normalized noise power spectrum is determined from a Monte Carlo analysis. This factor was calculated in four converter materials: amorphous silicon (a-Si), amorphous selenium (a-Se), cesium
Mowei Zhou et al.
Analytical chemistry, 84(14), 6016-6023 (2012-07-04)
A custom in-line surface-induced dissociation (SID) device has been incorporated into a commercial ion mobility quadrupole/time-of-flight mass spectrometer in order to provide an alternative and potentially more informative activation method than the commonly used collision-induced dissociation (CID). Complicated sample mixtures
Marco Bertolini et al.
Medical physics, 39(5), 2617-2627 (2012-05-09)
The purpose of this study is to compare digital radiography systems using the metric effective detective quantum efficiency (eDQE), which better reflects digital radiography imaging system performance under clinical operating conditions, in comparison with conventional metrics such as modulation transfer
Nobuhiro Yasui et al.
Advanced materials (Deerfield Beach, Fla.), 24(40), 5464-5469 (2012-08-14)
Alkali halide systems that function as phase-separated scintillators (PSSs) with light-guiding properties are sucessfully created. Furthermore, it is the matrix phases of the PSSs which display the light-guiding properties. CsI-NaCl:Tl is a practical material pair because of its high pixel
Melanie Freed et al.
Medical physics, 37(6), 2593-2605 (2010-07-17)
Accurate models of detector blur are crucial for performing meaningful optimizations of three-dimensional (3D) x-ray breast imaging systems as well as for developing reconstruction algorithms that faithfully reproduce the imaged object anatomy. So far, x-ray detector blur has either been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service