Skip to Content
Merck
All Photos(1)

Documents

429422

Sigma-Aldrich

Potassium iodide

AnhydroBeads, −10 mesh, 99.998% trace metals basis

Synonym(s):

Potassium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 745 °C)

product line

AnhydroBeads

Assay

99.998% trace metals basis

form

beads

impurities

<100 ppm water

particle size

−10 mesh

mp

681 °C (lit.)

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Clement et al.
Pediatric blood & cancer, 60(11), 1833-1838 (2013-07-09)
Thyroid dysfunction has been reported in up to 52% of patients 1.4 years after treatment with (131) I-Metaiodobenzylguanidine (MIBG) in children with neuroblastoma (NBL), despite the use of potassium-iodide (KI). Our aim was to investigate if the incidence and severity
J B Sterling et al.
Journal of the American Academy of Dermatology, 43(4), 691-697 (2000-09-27)
Potassium iodide (KI) is a useful drug in the dermatologic armamentarium. It is successfully used for inflammatory dermatoses, most notably erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet's syndrome. KI is also successfully used for cutaneous
Pietro Santulli et al.
Human reproduction (Oxford, England), 30(1), 49-60 (2014-11-08)
Are protein oxidative stress markers [thiols, advanced oxidation protein products (AOPP), protein carbonyls and nitrates/nitrites] in perioperative peritoneal fluid higher in women with histologically proven endometriosis when compared with endometriosis-free controls? Protein oxidative stress markers are significantly increased in peritoneal
Ilzira A Minigalieva et al.
International journal of molecular sciences, 16(9), 22555-22583 (2015-09-24)
Stable suspensions of NiO and Mn₃O₄ nanoparticles (NPs) with a mean (±s.d.) diameter of 16.7±8.2 and 18.4±5.4 nm, respectively, purposefully prepared by laser ablation of 99.99% pure nickel or manganese in de-ionized water, were repeatedly injected intraperitoneally (IP) to rats
Zhi-Qi Lao et al.
Organic & biomolecular chemistry, 10(39), 7869-7871 (2012-09-06)
We have developed a new KI-catalyzed method for the imidation of an sp(3) C-H bond adjacent to an amide nitrogen atom by using TBHP (tert-butyl hydroperoxide, 70% aqueous solution) as the oxidant. This novel procedure tolerated air and moisture and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service