Skip to Content
Merck
All Photos(1)

Key Documents

184063

Sigma-Aldrich

6-Methoxy-3,4-dihydro-2(1H)-naphthalenone

95%

Synonym(s):

6-Methoxy-2-tetralone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.5645 (lit.)

mp

30-37 °C

functional group

ketone

storage temp.

2-8°C

SMILES string

COc1ccc2CC(=O)CCc2c1

InChI

1S/C11H12O2/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h3,5,7H,2,4,6H2,1H3

InChI key

RMRKDYNVZWKAFP-UHFFFAOYSA-N

Related Categories

Application

6-Methoxy-3,4-dihydro-2(1H)-naphthalenone was used in the synthesis of 1, 2, 3, 4-tetrahydro-2-naphthylamine derivatives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kanao et al.
Journal of medicinal chemistry, 25(11), 1358-1363 (1982-11-01)
N-[(Benzoyloxy)alkyl]-1,2,3,4-tetrahydro-2-naphthylamine derivatives were synthesized from 1,2,3,4-tetrahydro-2-naphthylamines and evaluated for their spasmolytic. Some of these compounds showed a nerve-selective effect on colon rather than stomach in anesthetized dogs and were found to be equal to or more active than the reference

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service