Skip to Content
Merck
All Photos(1)

Key Documents

273937

Sigma-Aldrich

6,7-Dimethoxy-1-tetralone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

98-100 °C (lit.)

functional group

ketone

SMILES string

COc1cc2CCCC(=O)c2cc1OC

InChI

1S/C12H14O3/c1-14-11-6-8-4-3-5-10(13)9(8)7-12(11)15-2/h6-7H,3-5H2,1-2H3

InChI key

YNNJHKOXXBIJKK-UHFFFAOYSA-N

General description

6,7-Dimethoxy-1-tetralone reacts with 2-amino-4,5-dimethoxyacetophenone to form 5,6-dihydro-2,3,9,10-tetramethoxybenz[c]acridine.

Application

6,7-Dimethoxy-1-tetralone was used in the synthesis of 2-bromotetralones by undergoing bromination. It was also used as a precursor to quinolines with dopaminergic activity, naphthols with anti-inflammatory activity and benzophenanthridine alkaloids with antitumor activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin J Dunsmore et al.
Bioorganic & medicinal chemistry letters, 18(5), 1730-1734 (2008-02-12)
Several 2-aminotetralones were identified as novel inhibitors of the bacterial enzymes MurA and MurZ. A number of these inhibitors demonstrated antibacterial activity against Staphylococcus aureus and Escherichia coli with MICs in the range 8-128 microg/ml. Based on structure-activity relationships we
The Journal of Organic Chemistry, 57, 5907-5907 (1992)
D G Batt et al.
Journal of medicinal chemistry, 33(1), 360-370 (1990-01-01)
The synthesis, biological evaluation, and structure-activity relationships of a series of 1-naphthols bearing carbon substituents at the 2-position are described. These compounds are potent inhibitors of the 5-lipoxygenase from RBL-1 cells and also inhibit bovine seminal vesicle cyclooxygenase. Structure-activity relationships
J C Craig et al.
Journal of medicinal chemistry, 32(5), 961-968 (1989-05-01)
A series of 2-substituted octahydrobenzo[f]quinolines has been synthesized and assayed for dopamine agonist activity. Only the compounds corresponding to the beta-rotameric conformation of dopamine showed biphasic activity in competition binding studies with the radioligand [3H]spiroperidol. These findings suggest that the
D Makhey et al.
Bioorganic & medicinal chemistry, 8(5), 1171-1182 (2000-07-06)
Coralyne and several other synthetic benzo[a,g]quinolizium derivatives related to protoberberine alkaloids have exhibited activity as topoisomerase poisons. These compounds are characterized by the presence of a positively charged iminium group, which has been postulated to be associated with their pharmacological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service