708453
7-Bromo-3,4-dihydronaphthalen-1(2H)-one
97%
Synonym(s):
7-Bromo-1-tetralone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H9BrO
CAS Number:
Molecular Weight:
225.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
75-80 °C
functional group
bromo
ketone
storage temp.
2-8°C
SMILES string
Brc1ccc2CCCC(=O)c2c1
InChI
1S/C10H9BrO/c11-8-5-4-7-2-1-3-10(12)9(7)6-8/h4-6H,1-3H2
InChI key
YGVDCGFUUUJCDF-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service