Skip to Content
Merck
All Photos(1)

Key Documents

175420

Sigma-Aldrich

2,5-Dimethylpyrazine

98%

Synonym(s):

2,5-Dimethylpyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
107052
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.502 (lit.)

bp

155 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

Cc1cnc(C)cn1

InChI

1S/C6H8N2/c1-5-3-8-6(2)4-7-5/h3-4H,1-2H3

InChI key

LCZUOKDVTBMCMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Pickard et al.
Journal of agricultural and food chemistry, 61(26), 6274-6281 (2013-06-12)
A stable isotope dilution analysis based on gas chromatography-mass spectrometry analysis (SIDA-GC-MS) was developed for the quantitative analysis of 12 alkylpyrazines found in commercially available coffee samples. These compounds contribute to coffee flavor. The accuracy of this method was tested
Pyrazine and 2, 5-dimethylpyrazine complexes of copper (I) trifluoromethanesulfonate. The crystal and molecular structure of poly-μ-2, 5-dimethylpyrazine-(2, 5-dimethylpyrazine)-(trifluoromethanesulfonato-O) copper (I).
Canadian Journal of Chemistry, 68(10), 1901-1907 (1990)
David C Robacker et al.
Journal of chemical ecology, 35(5), 601-609 (2009-05-12)
Emissions from sexually active Anastrepha serpentina males were collected by solid-phase microextraction. Calling behavior of wild-type males showed no clear peak during the day, except that it was evident less frequently immediately after daybreak and just before dark. Calling by
K Yamada et al.
Biological & pharmaceutical bulletin, 24(9), 1068-1071 (2001-09-18)
The effect of alkylpyrazine derivatives on pentobarbital-induced sleeping time, picrotoxicin-induced convulsion and gamma-aminobutyric acid (GABA) levels in mouse brain were studied. The duration of pentobarbital-induced sleep in mice was dose-dependently increased by 2,5-dimethylpyrazine (DMP). The duration of pentobarbital-induced sleep was
K Yamada et al.
Biological & pharmaceutical bulletin, 17(5), 730-731 (1994-05-01)
The effects of 2,5-dimethylpyrazine (DMP) on plasma testosterone and levels of polyamines and acid phosphatase in the prostate of rat were studied. A high dose of DMP administered once daily for two weeks to juvenile male rats significantly decreased plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service