Skip to Content
Merck
All Photos(2)

Key Documents

95162

Supelco

2,3,5,6-Tetramethylpyrazine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
Beilstein:
113100
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

190 °C (lit.)

mp

77-80 °C (lit.)
85-87 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1nc(C)c(C)nc1C

InChI

1S/C8H12N2/c1-5-6(2)10-8(4)7(3)9-5/h1-4H3

InChI key

FINHMKGKINIASC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5,6-Tetramethylpyrazine is a flavor compound belonging to the class of pyrazines. It is found to be a main active ingredient of the traditional Chinese medicine, Chuanxiong and Ma-Huang herbs, used to treat asthma.

Application

2,3,5,6-Tetramethylpyrazine may be used as an analytical reference standard for the quantification of the analyte in Ephedrae herba and Ephedra sinica Stapf using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of tetramethylpyrazine in Ephedrae herba by gas chromatography?mass spectrometry and high-performance liquid chromatography
Lv K, et al.
Journal of Chromatography A, 878(1), 147-152 (2000)
Applications of Microbial Engineering (2013)
Separation and determination of ephedrine alkaloids and tetramethylpyrazine in ephedra sinica Stapf by gas chromatography-mass spectrometry
Journal of Chromatographic Science, 39(9), 370-374 (2001)
Shouliang Wang et al.
Drug design, development and therapy, 13, 1187-1196 (2019-05-23)
Background: Bupivacaine (BUP) acts as a local anesthetic, which is extensively used for clinical patients but could generate neurotoxicity in neurons. Tetramethylpyrazine (TET) exhibits strong neuron protective effects against neurotoxicity. Hence, we investigate the effect of TET on BUP-induced neurotoxicity
Lijun Wei et al.
International journal of pharmaceutics, 437(1-2), 203-212 (2012-09-05)
Ligustrazine is a traditional Chinese medicine used to treat various cardiovascular and neurovascular complications. However, this compound exhibits rapid first-pass metabolism, a short biological half-life, low stability and potential vascular irritation that restrict its use for long-term therapy. The use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service