Skip to Content
Merck
All Photos(1)

Key Documents

W328103

Sigma-Aldrich

2-Ethylpyrazine

≥98%, FG

Synonym(s):

Ethylpyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
FEMA Number:
3281
Beilstein:
108200
EC Number:
Council of Europe no.:
2213
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.022
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.498 (lit.)

bp

152-153 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cocoa; musty; nutty; woody; roasted

SMILES string

CCc1cnccn1

InChI

1S/C6H8N2/c1-2-6-5-7-3-4-8-6/h3-5H,2H2,1H3

InChI key

KVFIJIWMDBAGDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Ethylpyrazine is a volatile aroma compound that is mainly formed in food products such as roasted coffee beans or roasted sesame seeds due to the Maillard reaction between sugars and proteins during the roasting process.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.0 °F

Flash Point(C)

42.78 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Odor threshold levels of pyrazine compounds and assessment of their role in the flavor of roasted foods.
Koehler PE, et al.
Journal of Food Science, 36(5), 816-818 (1971)
Effect of antioxidants on the volatiles of roasted sesame seeds.
Soliman MA, et al.
Journal of Agricultural and Food Chemistry, 33(3), 523-528 (1985)
Steam volatile aroma constituents of roasted cocoa beans.
Van Praag M, et al.
Journal of Agricultural and Food Chemistry, 16(6), 1005-1008 (1968)
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service