Skip to Content
Merck
All Photos(3)

Key Documents

B59497

Sigma-Aldrich

1-Bromobutane

ReagentPlus®, 99%

Synonym(s):

Butyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Br
CAS Number:
Molecular Weight:
137.02
Beilstein:
1098260
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.7 (vs air)

Quality Level

vapor pressure

150 mmHg ( 50 °C)
40 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

509 °F

expl. lim.

2.8-6.6 %, 100 °F

refractive index

n20/D 1.439 (lit.)

bp

100-104 °C (lit.)

mp

−112 °C (lit.)

density

1.276 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCCCBr

InChI

1S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3

InChI key

MPPPKRYCTPRNTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Bromobutane, an alkyl halide, is an alkylating agent. Its rotational constants, nuclear quadrupole constants and centrifugal distortion constants have been stated based on microwave spectral data. The rate coefficient for the reaction of 1-bromobutane with hydrogen atoms has been reported to be 2.4±1.2x1010cm3mol-1s-1.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Liver, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rate coefficients for the reactions of hydrogen atoms with 1-bromopropane, 2-bromopropane, 1-bromobutane, and 2-bromobutane.
Meinike T, et al.
International Journal of Chemical Kinetics, 30(10), 721-727 (1998)
Francesco Giganti et al.
Radiology, 276(2), 444-452 (2015-03-31)
To prospectively investigate the role of apparent diffusion coefficient (ADC) calculated from diffusion-weighted magnetic resonance (MR) imaging as a potential prognostic biomarker in the evaluation of the aggressiveness of gastric cancer. This prospective study had institutional review board approval. Informed
W C E Schofield et al.
ACS applied materials & interfaces, 1(12), 2763-2767 (2010-04-02)
Existing methods for imparting antibacterial performance to solid surfaces tend to either be substrate-specific or rely upon leaching modes of action that cause ecological damage. An alternative approach is outlined comprising plasmachemical functionalization of solid surfaces with poly(4-vinyl pyridine) moieties
Sang Kyu Lee et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(10), 2707-2711 (2010-07-06)
Halogenated organic compounds, such as 1-bromobutane (1-BB), have been used as cleaning agents, agents for chemical syntheses, or extraction solvents. In the present study, hepatotoxic effects of 1-BB and its conjugation with glutathione (GSH) were investigated in female BALB/c mice.
Christina Lederle et al.
The Journal of chemical physics, 134(6), 064512-064512 (2011-02-17)
Mixtures of the monohydroxy alcohol n-butanol with n-bromobutane are investigated via dielectric and nuclear magnetic resonance (NMR) techniques. Static- and pulsed-field gradient proton NMR yielded self-diffusion coefficients as a function of concentration and temperature. To monitor reorientational motions, broadband dielectric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service