Merck
  • DNA-based mutation assay GPMA (genome profiling-based mutation assay): reproducibility, parts-per-billion scale sensitivity, and introduction of a mammalian-cell-based approach.
DNA-based mutation assay GPMA (genome profiling-based mutation assay): reproducibility, parts-per-billion scale sensitivity, and introduction of a mammalian-cell-based approach.
Journal of biochemistry (2017-12-01)
Parmila Kumari, Sunita Ghimire Gautam, Misato Baba, Motoki Tsukiashi, Koji Matsuoka, Kiyoshi Yasukawa, Koichi Nishigaki
ABSTRACT
Genome profiling-based mutation assay (GPMA) is, to date, the only DNA sequence-based mutation assay that directly measures DNA alterations induced by mutagens. Here, the all-important congruence of mutagen assignment between DNA-based GPMA and the phenotype-based Ames test (the gold standard of mutagen assays) was confirmed qualitatively and semi-quantitatively by means of 94 chemical species (including previously examined 64). The high sensitivity (on the order of 10 ppb) and reproducibility of GPMA were also corroborated by the match between virtually independent experiments conducted in the distant past (10 years ago) and recently. Meanwhile, a standard experimental framework was established: the conditions of 100 parts per billion (ppb) concentration of a chemical and 15-generation culture of Escherichia coli. Moreover, a mammalian cell line (NIH 3T3) was shown to be suitable as a tester organism for the GPMA approach. Preliminary experimental results suggested that this approach can provide a qualitatively equivalent and quantitatively different mutagen assay results relative to the bacteria-based GPMA (renamed as bGPMA). This finding confirmed the effectiveness of the GPMA approach and indicates that mGPMA is a promising way to detect mammalian-cell mutagens.
Sigma-Aldrich
1-Bromooctane, 99%