Merck
  • Changes in hypothalamic neurotransmitter and prostanoid levels in response to NMDA, CRF, and GLP-1 stimulation.
Changes in hypothalamic neurotransmitter and prostanoid levels in response to NMDA, CRF, and GLP-1 stimulation.
Analytical and bioanalytical chemistry (2015-01-31)
Fumio Kondo, Masahiko Tachi, Masahiko Gosho, Minoru Fukayama, Kazuhiro Yoshikawa, Shoshiro Okada
ABSTRACT
Determination of neuroactive substances, such as neurotransmitters and prostanoids, in the extracellular space of the living brain is a very important technique in neuroscience. The hypothalamic paraventricular nucleus (PVN) is one of the most important autonomic control centers in the brain. Recently, we demonstrated that thromboxane (Tx) A2 in the PVN may play an important role in adrenomedullary outflow evoked by N-methyl-D-aspartate (NMDA), corticotrophin-releasing factor (CRF), and glucagon-like peptide-1 (GLP-1) stimulation using microdialysis sampling and liquid chromatography-ion trap tandem mass spectrometry (LC-ITMS(n)). In the present study, we investigated whether centrally administered NMDA, CRF, and GLP-1 can release five neurotransmitters, acetylcholine (ACh), histamine, glutamate (Glu), γ-aminobutyric acid (GABA), and serotonin (5-HT), along with six prostanoids, TxB2, prostaglandin (PG) E2, PGD2, 15-deoxy-∆(12,14) (15d)-PGJ2, 6-keto-PGF1α, and PGF2α in rat PVN microdialysates after optimization of LC-ITMS(n) conditions . All stimulations increased the levels of 5-HT, TxB2, PGE2, and PGF2α (except for 5-HT stimulated with GLP-1). Only NMDA increased the levels of ACh, Glu, and GABA. Treatment with dizocilpine maleate (MK-801), a noncompetitive NMDA receptor antagonist, attenuated the NMDA-induced increase in the levels of neuroactive substances except for Glu. This is the first study to use LC-ITMS(n) to analyze neurotransmitters and prostanoids in the same microdialysates from rat PVN.
Sigma-Aldrich
Benzoyl chloride-d5, 99 atom % D
Sigma-Aldrich
Benzoyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzoyl chloride, 99%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methyl acetate, ≥98%, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Methyl acetate, anhydrous, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium tetraborate, 99%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate