Skip to Content
Merck
All Photos(3)

Documents

229946

Sigma-Aldrich

Sodium tetraborate

99.998% trace metals basis

Synonym(s):

Borax, fused

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7
CAS Number:
Molecular Weight:
201.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

99.998% trace metals basis

form

powder

mp

741 °C (lit.)

density

2.367 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1

InChI key

UQGFMSUEHSUPRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium tetraborate, also known as borax(Na2B4O7), can be employed as a catalyst:
  • In the chemoselective oxidation of alkyl and aryl sulfides to sulfoxides and sulfones using H2O2 as the oxidant.
  • In the synthesis of pyridines, dienes, anilines, and dihydropyrano[3,2-c]chromenes via Knoevenagel condensation and Michael addition pathway.
  • In thiolysis of 1,2-epoxides to β-hydroxy sulfides.

Legal Information

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Borax-Catalyzed and pH-Controlled Selective Oxidation of Organic Sulfides by H2O2: An Environmentally Clean Protocol
Hussain S, et al.
European Journal of Organic Chemistry, 2009(20), 3319-3322 (2009)
Borax-catalyzed thiolysis of 1, 2-epoxides in aqueous medium
Gao P, et al.
Tetrahedron Letters, 49(46), 6536-6538 (2008)
Borax catalyzed domino reactions: synthesis of highly functionalised pyridines, dienes, anilines and dihydropyrano [3, 2-c] chromenes
Molla A and Hussain S
Royal Society of Chemistry Advances, 4(56), 29750-29758 (2014)
Shuai Liu et al.
Journal of the American Heart Association, 9(8), e014757-e014757 (2020-04-21)
Background The protective effects of polyamines on cardiovascular disease have been demonstrated in many studies. However, the roles of spermidine, a natural polyamine, in abdominal aortic aneurysm (AAA) disease have not been studied. In this study, we investigated the influence
Shridhar K Sathe et al.
Journal of agricultural and food chemistry, 57(17), 7846-7856 (2009-08-07)
The solubility of almond, Brazil nut, cashew nut, hazelnut, macadamia, pecan, pine nut, pistachio, walnut, and peanut proteins in several aqueous solvents was qualitatively and quantitatively assessed. In addition, the effects of extraction time and ionic strength on protein solubility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service