Merck
  • Transbuccal delivery of doxepin: studies on permeation and histological investigation.
Transbuccal delivery of doxepin: studies on permeation and histological investigation.
International journal of pharmaceutics (2014-12-03)
Alvaro Gimeno, Ana C Calpena, Roser Sanz, Mireia Mallandrich, Concepción Peraire, Beatriz Clares
ABSTRACT
According to previous studies reporting the anesthetic/analgesic action of oral topical doxepin administration, this study evaluated a model of buccal permeation to determine the depth of delivery of doxepin into excised porcine buccal mucosa following topical application of a saturated aqueous doxepin solution. Buccal mucosa permeation studies were performed using Franz diffusion cells. Cumulative amounts of doxepin permeated were plotted as a function of time. Kinetic permeation parameters as flux (Js), lag time (Tl) and permeability coefficient (Kp) were calculated. Theoretical human plasmatic steady-state doxepin concentration and drug retained in the tissue were also determined in order to evaluate its potential therapeutic use, central or peripheral. Finally, a histological evaluation of the buccal mucosa was performed to test potential damage due to the permeation phenomenon. Obtained results showed a poor aqueous doxepin permeation through buccal mucosa membrane (median parameters Js=34.79 μg/h, Kp=0.49×10(-3)cm/h and Tl=2.8h). Predicted doxepin plasma concentrations would reach 46 ng/mL, far from the required to have central nervous system activity as tricyclic agent. However, median doxepin amount remaining in the mucosa membrane was 0.24 μg/cm(2)/μg tissue, which evidenced a reservoir function of the buccal mucosa. Histologically, no structural damage was observed in the tissues. This study lays the foundation for further research within this area with a view to potentially adopting alternative strategies for enhanced buccal absorption of doxepin in clinical practice.
Product Description
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Doxepin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Doxepin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)