Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

Merck

Key Documents

Skip To

D4526

Doxepin hydrochloride

≥98% (GC), norepinephrine uptake inhibitor, powder

Synonym(s):

11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenz[b,e]oxepine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
214-966-8
MDL number:
Assay:
≥98% (GC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder

Quality Segment

assay

≥98% (GC)

form

powder

composition

Z-isomer, 15% , E-isomer, ~85%

color

white to off-white

solubility

1 M HCl: 50 mg/mL, H2O: soluble

originator

Boehringer Ingelheim

SMILES string

Cl[H].CN(C)CC\C=C1/c2ccccc2COc3ccccc13

InChI

1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11+;

InChI key

MHNSPTUQQIYJOT-SJDTYFKWSA-N

General description

Doxepin is used to treat chronic idiopathic urticaria, chronic pain and anxiety. Doxepin exhibits antiserotonergic properties.

Biochem/physiol Actions

Tricyclic antidepressant that is a more potent inhibitor of norepinephrine uptake than of serotonin uptake; antagonist at H1 histamine, muscarinic cholinergic, and α-adrenoreceptors.

Features and Benefits

This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Mixture of cis- and trans-isomers

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP823Y0000851Y0000838
description

~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder

description

British Pharmacopoeia (BP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

assay

≥98% (GC)

assay

-

assay

-

assay

-

form

powder

form

solid

form

-

form

-

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

-

solubility

H2O: soluble, 1 M HCl: 50 mg/mL

solubility

-

solubility

-

solubility

-

originator

Boehringer Ingelheim

originator

-

originator

-

originator

-

color

white to off-white

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Doxepin hydrochloride


Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters