Merck
  • Susceptibility of clinically important dermatophytes against statins and different statin-antifungal combinations.
Susceptibility of clinically important dermatophytes against statins and different statin-antifungal combinations.
Medical mycology (2013-09-06)
Ildikó Nyilasi, Sándor Kocsubé, Krisztina Krizsán, László Galgóczy, Tamás Papp, Miklós Pesti, Katalin Nagy, Csaba Vágvölgyi
ABSTRACT
The investigation of the antifungal activities of drugs whose primary activities are not related to their antimicrobial potential is in the current forefront of research. Statin compounds, which are routinely used as cholesterol-lowering drugs, may also exert direct antimicrobial effects. In this study, the in vitro antifungal activities of various statins (lovastatin, simvastatin, fluvastatin, atorvastatin, rosuvastatin and pravastatin) were examined against one isolate each of four dermatophyte species (Trichophyton mentagrophytes, Trichophyton rubrum, Microsporum canis and Microsporum gypseum). Basically, statins were effective in inhibiting all dermatophyte studied, but were particularly active against M. canis and T. mentagrophytes. Fluvastatin and simvastatin were active against all of the tested fungi causing a complete inhibition of their growth at very low concentrations (6.25-12.5 μg/ml). Lovastatin and rosuvastatin had inhibitory effects at higher concentrations (25-128 μg/ml), while atorvastatin and pravastatin proved the less effective. The in vitro interactions between statins and different antifungals (ketoconazole, itraconazole, fluconazole, amphotericin B, nystatin, griseofulvin, terbinafine and primycin) were also investigated using a standard chequerboard broth microdilution method. Synergetic interactions were observed in several cases, most of them were noticed when statins were combined with terbinafine and the different azoles. Some combinations were particularly active (ketoconazole-simvastatin or terbinafine-simvastatin), as they were found to exert synergistic effect against all of the investigated isolates. The other antifungals showed synergistic interactions with statins in only certain cases. These results suggest that statins exert substantial antifungal effects against dermatophyte fungi and they should be promising components in a combination therapy as they can act synergistically with a number of clinically used antifungal agents.
Sigma-Aldrich
L-Glutamine
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Itraconazole, ≥98% (HPLC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Griseofulvin Permeability Diameter, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1-Propanesulfonic acid, technical, ≥99.0% (T)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Itraconazole, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard