Skip to Content
Merck
All Photos(1)

Key Documents

Y0000005

Amphotericin B

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

amphotericin b

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

N[C@@H]1[C@@H]([C@@H](O[C@@H]([C@H]1O)C)O[C@@H]2C[C@@H]3O[C@@](C[C@@H]([C@H]3C(=O)O)O)(C[C@H](C[C@H]([C@@H](CC[C@H](C[C@H](CC(=O)O[C@H]([C@@H]([C@@H]([C@H](\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)C)O)C)C)O)O)O)O)O)O)O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5-,9-7-,10-8-,13-11-,14-12-,17-15-,18-16-/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-ZNVUZQDLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amphotericin B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Bessède et al.
Oncogene, 33(32), 4123-4131 (2013-10-08)
Helicobacter pylori infection is the major risk factor for gastric adenocarcinoma. The link with gastric adenocarcinoma is partly due to the H. pylori CagA oncoprotein. CagA is responsible for a particular cell phenotype in vitro, the 'hummingbird' phenotype, that corresponds
Andrew B Allison et al.
PLoS pathogens, 10(11), e1004475-e1004475 (2014-11-07)
Canine parvovirus (CPV) emerged as a new pandemic pathogen of dogs in the 1970s and is closely related to feline panleukopenia virus (FPV), a parvovirus of cats and related carnivores. Although both viruses have wide host ranges, analysis of viral
Azadeh Hosseini-Tabatabaei et al.
Canadian journal of diabetes, 37(1), 27-35 (2013-09-28)
Islet transplantation is a promising strategy to restore efficient insulin regulation in type 1 diabetes mellitus patients. However, shortage of islet donors, poor islet survival and toxicity of immunosuppressants often reduce the graft functional lifetime. We previously showed that a
Manica Balasegaram et al.
Expert opinion on emerging drugs, 17(4), 493-510 (2012-11-22)
Leishmaniasis is a parasitic disease transmitted by phlebotomine sandflies. Between 700,000 and 1.2 million cases of cutaneous leishmaniasis and between 200,000 and 400,000 cases of visceral leishmaniasis (VL), which is fatal if left untreated, occur annually worldwide. Liposomal amphotericin B
David R Boulware et al.
The New England journal of medicine, 370(26), 2487-2498 (2014-06-26)
Cryptococcal meningitis accounts for 20 to 25% of acquired immunodeficiency syndrome-related deaths in Africa. Antiretroviral therapy (ART) is essential for survival; however, the question of when ART should be initiated after diagnosis of cryptococcal meningitis remains unanswered. We assessed survival

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service