Merck
Enzyme enhancers for the treatment of Fabry and Pompe disease.
Molecular therapy : the journal of the American Society of Gene Therapy (2014-11-21)
Jan Lukas, Anne-Marie Pockrandt, Susanne Seemann, Muhammad Sharif, Franziska Runge, Susann Pohlers, Chaonan Zheng, Anne Gläser, Matthias Beller, Arndt Rolfs, Anne-Katrin Giese
ABSTRACT
Lysosomal storage disorders (LSD) are a group of heterogeneous diseases caused by compromised enzyme function leading to multiple organ failure. Therapeutic approaches involve enzyme replacement (ERT), which is effective for a substantial fraction of patients. However, there are still concerns about a number of issues including tissue penetrance, generation of host antibodies against the therapeutic enzyme, and financial aspects, which render this therapy suboptimal for many cases. Treatment with pharmacological chaperones (PC) was recognized as a possible alternative to ERT, because a great number of mutations do not completely abolish enzyme function, but rather trigger degradation in the endoplasmic reticulum. The theory behind PC is that they can stabilize enzymes with remaining function, avoid degradation and thereby ameliorate disease symptoms. We tested several compounds in order to identify novel small molecules that prevent premature degradation of the mutant lysosomal enzymes α-galactosidase A (for Fabry disease (FD)) and acid α-glucosidase (GAA) (for Pompe disease (PD)). We discovered that the expectorant Ambroxol when used in conjunction with known PC resulted in a significant enhancement of mutant α-galactosidase A and GAA activities. Rosiglitazone was effective on α-galactosidase A either as a monotherapy or when administered in combination with the PC 1-deoxygalactonojirimycin. We therefore propose both drugs as potential enhancers of pharmacological chaperones in FD and PD to improve current treatment strategies.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Ambroxol hydrochloride, analytical standard
Ambroxol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Bezafibrate, ≥98%, solid
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Bezafibrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid