Skip to Content
Merck
All Photos(5)

Key Documents

D8284

Sigma-Aldrich

Bicinchoninic acid disodium salt hydrate

≥98% purity (HPLC), powder

Synonym(s):

2,2′-Biquinoline-4,4′-dicarboxylic acid disodium salt, 4,4′-Dicarboxy-2,2′-biquinoline disodium salt, BCA, Disodium 2,2′-biquinoline-4,4′-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10N2Na2O4 · xH2O
CAS Number:
Molecular Weight:
388.28 (anhydrous basis)
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

titration: suitable

color

white to light yellow

solubility

0.5% sodium bicarbonate: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].[O-]C(=O)c1cc(nc2ccccc12)-c3cc(C([O-])=O)c4ccccc4n3

InChI

1S/C20H12N2O4.2Na.H2O/c23-19(24)13-9-17(21-15-7-3-1-5-11(13)15)18-10-14(20(25)26)12-6-2-4-8-16(12)22-18;;;/h1-10H,(H,23,24)(H,25,26);;;1H2/q;2*+1;/p-2

InChI key

FAFVGCAYKZKOPT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Bicinchoninic acid disodium salt hydrate has been used to monitor the levels of reducing sugars, thereby helping in the detection of chitinases.[1]
  • It has also been used to determine the amount of cellulose reducing ends (micromoles/g cellulose).[2]
  • It has been used for determining protein concentration.[3]

Biochem/physiol Actions

Bicinchoninic acid is used for the determination of protein concentration. It is responsible for the making of intense purple complex with cuprous ion (Cu+), in presence of alkaline conditions. The amount of Cu+ is in relation with the protein concentration.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prevention of rt-PA induced blood-brain barrier component degradation by the poly(ADP-ribose)polymerase inhibitor PJ34 after ischemic stroke in mice.
Teng F, et al.
Experimental Neurology, 248, 416-428 (2013)
Detection and characterization of chitinases and other chitin-modifying enzymes.
Howard MB, et al.
Journal of Industrial Microbiology & Biotechnology, 30, 627-635 (2003)
Ramya Kumareswaran et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 116(3), 388-391 (2015-04-07)
Solid tumors contain regions of chronic and cyclic hypoxia. Chronic hypoxia can downregulate RAD51 and sensitize cells to PARP inhibition. Herein, we show that RAD51 expression, cell survival and toxicity to PARP inhibition is not affected under cyclic hypoxic conditions.
Makon-Sébastien Njock et al.
Blood, 125(20), 3202-3212 (2015-04-04)
The blood contains high concentrations of circulating extracellular vesicles (EVs), and their levels and contents are altered in several disease states, including cardiovascular disease. However, the function of circulating EVs, especially the microRNAs (miRNAs) that they contain, are poorly understood.
Rajeev Kumar et al.
Bioresource technology, 130, 372-381 (2013-01-15)
Two established delignification methods employing sodium chlorite-acetic acid (SC/AA) and peracetic acid (PAA) are often used, and are reportedly highly selective. However, these reports are mostly for highly recalcitrant and unpretreated softwoods and hardwoods species, and information for less recalcitrant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service